EMAIL THIS PAGE TO A FRIEND
The American journal of the medical sciences
Synergistic Effects of Combining Anti-Midkine and Hepatocyte Growth Factor Therapies Against Diabetic Nephropathy in Rats.
PMID 26086153
Abstract
This study aimed to assess whether synergism could be achieved when combining midkine (MK) antisense oligodeoxynucleotides (anti-MK ODN) and recombinant human hepatocyte growth factor (HGF) in diabetic nephropathy (DN) rat models. Rats were randomized into 6 groups: control, DN rats without treatment, DN rats treated with scrambled ODN, DN rats treated with anti-MK ODN, DN rats treated with HGF and DN rats treated with anti-MK ODN plus HGF. DN models were created by intraperitoneal injection of streptozotocin. Two weeks later, treatments commenced. ODN (1 mg/kg) was intravenously injected weekly for 4 weeks. HGF (500 μg/kg) was subcutaneously injected daily for 4 weeks. Eight weeks later, rats were euthanized. Serum and urine parameters, kidney histopathological injury scores, immunohistochemistry and protein expressions were measured. Blood glucose, creatinine, blood urea nitrogen and urine albumin were significantly elevated in DN rats. Any single treatment markedly reduced their levels, yet combined treatment decreased them significantly further. Any monotherapy could decrease renal injury score and immunohistochemistry positive percentage, although the most prominent change was displayed in combinational therapy. Western blot showed the expression of MK was significantly elevated in DN rats. Anti-MK ODN suppressed MK significantly. The protein expressions and serum concentrations of transforming growth factor-β1 and connective tissue growth factor between monotherapy and the combined therapy were significant. This study demonstrated that combining MK gene suppressing ODN and HGF protein synergistically attenuates renal injury in DN rats. This study may provide a novel avenue for designing future therapeutic regimens against DN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21023 Anti-ATP7A antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA012887
Anti-ATP7A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33797
Anti-ATP7A antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2104637
Anti-ATP7A antibody produced in rabbit, affinity isolated antibody
SAB3500427
Anti-DR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1300911
Anti-MDK (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1406113
Anti-MDK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409304
Anti-MDK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410778
Anti-MDK antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501935
Anti-Mevalonate Kinase antibody produced in rabbit, affinity isolated antibody
SAB1406150
Anti-MVK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA016961
Anti-MVK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401251
Anti-MVK antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2104112
Anti-MVK antibody produced in rabbit, affinity isolated antibody
SAB2104111
Anti-MVK, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1406725
Anti-TNFRSF10B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA023625
Anti-TNFRSF10B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102489
Anti-TNFRSF10B antibody produced in rabbit, affinity isolated antibody
H9661 Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
H5791 Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
SRP6014 HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
SRP6166
HGF human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SAB4700538 Monoclonal Anti-CD262 antibody produced in mouse, clone DR5-01-1, purified immunoglobulin, buffered aqueous solution
SAB1402278
Monoclonal Anti-MVK, (C-terminal) antibody produced in mouse, clone 2C5, purified immunoglobulin, buffered aqueous solution
WH0008795M1
Monoclonal Anti-TNFRSF10B antibody produced in mouse, clone 2D6, purified immunoglobulin, buffered aqueous solution