EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Advances in Use of Capsule-Based Fluorescent Sensors for Measuring Acidification of Endocytic Compartments in Cells with Altered Expression of V-ATPase Subunit V1G1.
PMID 26086317
Abstract
Acidification of eukaryotic cell compartments is accomplished by vacuolar H+-ATPases (V-ATPases), large multisubunit complexes able to pump protons into the lumen of organelles or in the extracellular medium. V-ATPases are involved in a number of physiological cellular processes, and thus regulation of V-ATPase activity is of crucial importance for the cell. Indeed, dysfunction of V-ATPase or alterations of acidification have been recently recognized as key factors in a variety of human diseases. In this study, we applied capsule-based pH sensors and a real-time tracking method for investigating the role of the V1G1 subunit of V-ATPases in regulating the activity of the proton pump. We first constructed stable cell lines overexpressing or silencing the subunit V1G1. Second, we used fluorescent capsule-based pH sensors to monitor acidification before and during internalization by modified and control living cells. By using a simple real-time method for tracking capsule internalization, we were able to identify different capsule acidification levels with respect to each analyzed cell and to establish the kinetics for each. The intracellular pH measurements indicate a delay in acidification in either V1G1-overexpressing or V1G1-silenced cells compared to controls. Finally, in an independent set of experiments, we applied transmission electron microscopy and confocal fluorescence microscopy to further investigate the internalization of the capsules. Both analyses confirm that capsules are engulfed in acidic vesicular structures in modified and control cell lines. The use of capsule-based pH sensors allowed demonstration of the importance of the V1G1 subunit in V-ATPase activity concerning intravesicular acidification. We believe that the combined use of these pH-sensor system and such a real-time method for tracking their internalization path would contribute to systematically measure the proton concentration changes inside the endocytic compartments in various cell systems. This approach would provide fundamental information regarding molecular mechanisms and factors that regulate intracellular acidification, vesicular trafficking, and cytoskeletal reorganizations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
186260
Ethanesulfonic acid, 95%
C2H6O3S
471550
Ethanesulfonic acid solution, 70 wt. % in H2O
C2H6O3S
E5134
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration)
C10H14N2Na2O8 · 2H2O
E1644
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E6635
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, plant cell culture tested, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E5513
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E4884
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
ED2SS
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
C10H14N2Na2O8 · 2H2O
27285
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
G7513
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
P4333
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, sterile-filtered, BioReagent, suitable for cell culture
283924
Rhodamine B isothiocyanate, mixed isomers
C29H30ClN3O3S
R1755
Rhodamine B isothiocyanate, BioReagent, mixed isomers, suitable for protein labeling
C29H30ClN3O3S