EMAIL THIS PAGE TO A FRIEND
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society
Lactobacillus rhamnosus GG supernatant upregulates serotonin transporter expression in intestinal epithelial cells and mice intestinal tissues.
PMID 26088715
Abstract
The role that probiotics play in relieving irritable bowel syndrome (IBS) has been demonstrated; however, the mechanism by which IBS is affected remains unclear. In this study, serotonin transporter (SERT) mRNA and serotonin transporter protein (SERT-P) levels in HT-29, Caco-2 cells, and mice intestinal tissues were examined after treatment with Lactobacillus rhamnosus GG supernatant (LGG-s). HT-29 and Caco-2 cells were treated with different concentrations of LGG-s for 12 and 24xa0h and C57BL/6 mice received supplements of different concentrations for 4xa0weeks. SERT mRNA and SERT-P levels were detected by real-time PCR and Western blotting. SERT mRNA and SERT-P levels in HT-29 and Caco-2 cells were higher than those in the control 24xa0h after treatment. Undiluted LGG-s upregulated SERT mRNA levels by 9.4-fold in the first week, which dropped in the second week. The double-diluted LGG-s upregulated SERT mRNA by 2.07-fold in the first week; levels dropped to 1.75-fold within the second week and under base expression levels by the third week, while they again climbed to 1.56-fold in the fourth week. The triple-diluted LGG-s could not upregulate SERT mRNA expression until the end of the fourth week. The SERT-P levels in the double-diluted LGG-s group were higher than that in the control but fluctuated with time. SERT-P levels in the triple-diluted LGG-s were higher than that in the control in the last 2xa0weeks and increased with time. LGG-s can upregulate SERT mRNA and SERT-P levels in intestinal epithelial cells and mice intestinal tissues.