EMAIL THIS PAGE TO A FRIEND
BioMed research international
Bone Marrow Cells in Acute Lymphoblastic Leukemia Create a Proinflammatory Microenvironment Influencing Normal Hematopoietic Differentiation Fates.
PMID 26090405
Abstract
B-cell acute lymphoblastic leukemia (B-ALL) is a serious public health problem in the pediatric population worldwide, contributing to 85% of deaths from childhood cancers. Understanding the biology of the disease is crucial for its clinical management and the development of therapeutic strategies. In line with that observed in other malignancies, chronic inflammation may contribute to a tumor microenvironment resulting in the damage of normal processes, concomitant to development and maintenance of neoplastic cells. We report here that hematopoietic cells from bone marrow B-ALL have the ability to produce proinflammatory and growth factors, including TNFα, IL-1β, IL-12, and GM-CSF that stimulate proliferation and differentiation of normal stem and progenitor cells. Our findings suggest an apparently distinct CD13(+)CD33(+) population of leukemic cells contributing to a proinflammatory microenvironment that may be detrimental to long-term normal hematopoiesis within B-ALL bone marrow.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21888
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C29H19NO11
87444
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
C29H19NO11
GW10811F Anti-CD34 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300690
Anti-CD34 antibody produced in rabbit, affinity isolated antibody
SAB4300034
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
SAB4504540
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
S2690 Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300033
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
SAB4504541
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
S5386
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
C3H7NO3
S4375
DL-Serine, ≥98% (TLC)
C3H7NO3
145726
DL-Tyrosine, 99%
C9H11NO3
WH0000947M1
Monoclonal Anti-CD34 antibody produced in mouse, clone 5F3, purified immunoglobulin, buffered aqueous solution
SAB1403649
Monoclonal Anti-CD34 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
SAB4700160 Monoclonal Anti-CD34 antibody produced in mouse, clone 4H11[APG], purified immunoglobulin, buffered aqueous solution
SAB4700736 Monoclonal Anti-CD34 antibody produced in mouse, clone 581, purified immunoglobulin, buffered aqueous solution
PZ0203
Sitaxentan sodium salt, ≥98% (HPLC)
C18H14ClN2NaO6S2