EMAIL THIS PAGE TO A FRIEND
Journal of immunology research
Umbilical Cord-Derived Mesenchymal Stem Cells Inhibit Cadherin-11 Expression by Fibroblast-Like Synoviocytes in Rheumatoid Arthritis.
PMID 26090476
Abstract
This study aimed to determine whether umbilical cord-derived mesenchymal stem cells (UCMSC) regulate Cadherin-11 (CDH11) expression by fibroblast-like synoviocytes (FLS) in rheumatoid arthritis (RA). FLS were isolated from the synovium of RA and osteoarthritis (OA) patients. FLS from RA patients were cocultured with UCMSC in a transwell system. CDH11 mRNA levels in FLS were tested, and levels of soluble factors expressed by UCMSC, such as indoleamine 2,3-dioxygenase (IDO), hepatocyte growth factor (HGF), and interleukin- (IL-) 10, were determined. IDO, HGF, and IL-10 were upregulated in cocultures, so that appropriate inhibitors were added before determination of CDH11 expression. The effects of UCMSC on arthritis were investigated in the collagen-induced arthritis (CIA) model in Wistar rats. FLS from RA patients expressed higher CDH11 levels than those from OA patients, and this effect was suppressed by UCMSC. The inhibitory effect of UCMSC on CDH11 expression by FLS was abolished by suppression of IL-10 activity. CDH11 expression in synovial tissues was higher in the context of CIA than under basal conditions, and this effect was prevented by UCMSC administration. IL-10 mediates the inhibitory effect of UCMSC on CDH11 expression by FLS, and this mechanism might be targeted to ameliorate arthritis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501745
Anti-CDH11 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501326
Anti-CDH11 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4500033
Anti-CDH11 antibody produced in rabbit, affinity isolated antibody
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1409249
Anti-IL10 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410712
Anti-IL10 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
I5020 Anti-Interleukin 10 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
GW21031 Anti-Interleukin-10 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I5145 Anti-Interleukin-10 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
SAB4501868
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501869
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1306038
ANTI-TGIF (CENTER L223) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21970F Anti-TGIF antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
WH0001009M1
Monoclonal Anti-CDH11 antibody produced in mouse, clone 4D10, purified immunoglobulin, buffered aqueous solution
H1896 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24612.111, purified immunoglobulin, lyophilized powder