EMAIL THIS PAGE TO A FRIEND
Human vaccines & immunotherapeutics
H7N9 T-cell epitopes that mimic human sequences are less immunogenic and may induce Treg-mediated tolerance.
PMID 26090577
Abstract
Avian-origin H7N9 influenza is a novel influenza A virus (IAV) that emerged in humans in China in 2013. Using immunoinformatics tools, we identified several H7N9 T cell epitopes with T cell receptor (TCR)-facing residues identical to those of multiple epitopes from human proteins. We hypothesized that host tolerance to these peptides may impair T helper response and contribute to the low titer, weak hemagglutination inhibiting (HI) antibody responses and diminished seroconversion rates that have been observed in human H7N9 infections and vaccine trials. We found that the magnitude of human T effector responses to individual H7N9 peptides was inversely correlated with the peptide's resemblance to self. Furthermore, a promiscuous T cell epitope from the hemagglutinin (HA) protein suppressed responses to other H7N9 peptides when co-administered in vitro. Along with other highly 'human-like' peptides from H7N9, this peptide was also shown to expand FoxP3(+) regulatory T cells (Tregs). Thus, H7N9 may be camouflaged from effective human immune response by T cell epitope sequences that avert or regulate effector T cell responses through host tolerance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405588
Anti-ENTPD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014067
Anti-ENTPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503659
Anti-ENTPD1 antibody produced in rabbit, affinity isolated antibody
SAB2107618
Anti-ENTPD1 antibody produced in rabbit, affinity isolated antibody
AV41764
Anti-GNAS (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV41765
Anti-GNAS (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV42694
Anti-GNAS (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB2501411
Anti-GNAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA018122
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100941
Anti-GNAS antibody produced in rabbit, affinity isolated antibody
HPA027478
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028386
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB1405944
Anti-HLA-DRA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409196
Anti-HLA-DRA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410634
Anti-HLA-DRA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1400841
Anti-LIPH antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA049079
Anti-LIPH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107647
Anti-LIPH antibody produced in rabbit, affinity isolated antibody
HPA021656
Anti-SLC10A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105728
Anti-SLC10A3 antibody produced in rabbit, affinity isolated antibody