PloS one
SERR Spectroelectrochemical Study of Cytochrome cd1 Nitrite Reductase Co-Immobilized with Physiological Redox Partner Cytochrome c552 on Biocompatible Metal Electrodes.
PMID 26091174
Abstract
Cytochrome cd1 nitrite reductases (cd1NiRs) catalyze the one-electron reduction of nitrite to nitric oxide. Due to their catalytic reaction, cd1NiRs are regarded as promising components for biosensing, bioremediation and biotechnological applications. Motivated by earlier findings that catalytic activity of cd1NiR from Marinobacter hydrocarbonoclasticus (Mhcd1) depends on the presence of its physiological redox partner, cytochrome c552 (cyt c552), we show here a detailed surface enhanced resonance Raman characterization of Mhcd1 and cyt c552 attached to biocompatible electrodes in conditions which allow direct electron transfer between the conducting support and immobilized proteins. Mhcd1 and cyt c552 are co-immobilized on silver electrodes coated with self-assembled monolayers (SAMs) and the electrocatalytic activity of Ag // SAM // Mhcd1 // cyt c552 and Ag // SAM // cyt c552 // Mhcd1 constructs is tested in the presence of nitrite. Simultaneous evaluation of structural and thermodynamic properties of the immobilized proteins reveals that cyt c552 retains its native properties, while the redox potential of apparently intact Mhcd1 undergoes a ~150 mV negative shift upon adsorption. Neither of the immobilization strategies results in an active Mhcd1, reinforcing the idea that subtle and very specific interactions between Mhcd1 and cyt c552 govern efficient intermolecular electron transfer and catalytic activity of Mhcd1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674397
11-Amino-1-undecanethiol hydrochloride, 99%
C11H25NS · HCl
733679
6-Amino-1-hexanethiol hydrochloride
C6H15NS · HCl