PloS one
Destructive Changes in the Neuronal Structure of the FVB/N Mouse Retina.
PMID 26091175
Abstract
We applied a series of selective antibodies for labeling the various cell types in the mammalian retina. These were used to identify the progressive loss of neurons in the FVB/N mouse, a model of early onset retinal degeneration produced by a mutation in the pde6b gene. The immunocytochemical studies, together with electroretinogram (ERG) recordings, enabled us to examine the time course of the degenerative changes that extended from the photoreceptors to the ganglion cells at the proximal end of the retina. Our study indicates that photoreceptors in FVB/N undergo a rapid degeneration within three postnatal weeks, and that there is a concomitant loss of retinal neurons in the inner nuclear layer. Although the loss of rods was detected at an earlier age during which time M- and S-opsin molecules were translocated to the cone nuclei; by 6 months all cones had also degenerated. Neuronal remodeling was also seen in the second-order neurons with horizontal cells sprouting processes proximally and dendritic retraction in rod-driven bipolar cells. Interestingly, the morphology of cone-driven bipolar cells were affected less by the disease process. The cellular structure of inner retinal neurons, i.e., ChAT amacrine cells, ganglion cells, and melanopsin-positive ganglion cells did not exhibit any gross changes of cell densities and appeared to be relatively unaffected by the massive photoreceptor degeneration in the distal retina. However, Muller cell processes began to express GFAP at their endfeet at p14, and it climbed progressively to the cell's distal ends by 6 months. Our study indicates that FVB/N mouse provides a useful model with which to assess possible intervention strategies to arrest photoreceptor death in related diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2502122
Anti-CALB1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1408554
Anti-CALB1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501501
Anti-FOXC1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412610
ANTI-FOXC1 antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution
AV32300
Anti-FOXC1 antibody produced in rabbit, IgG fraction of antiserum
HPA040670
Anti-FOXC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500920
Anti-FOXC1 antibody produced in rabbit, affinity isolated antibody
SAB2104365
Anti-FOXC1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2105271
Anti-FOXC1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2500410
Anti-FOXC1, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1300371
Anti-PKCγ (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104731
Anti-PRKCG antibody produced in rabbit, affinity isolated antibody
SAB2104573
Anti-PRKCG, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1403824
Monoclonal Anti-FOXC1, (C-terminal) antibody produced in mouse, clone 4D11, purified immunoglobulin, buffered aqueous solution
SAB5200035
Monoclonal Anti-HCN4 antibody produced in mouse, clone S114-10, 1 mg/mL, purified immunoglobulin