EMAIL THIS PAGE TO A FRIEND
Lymphatic research and biology
A High-Salt Diet Differentially Modulates Mechanical Activity of Afferent and Efferent Collecting Lymphatics in Murine Iliac Lymph Nodes.
PMID 26091404
Abstract
The lymphatic system contributes to fluid homeostasis in various tissues. Recent evidence suggests that lymphangiogenesis induced by a high-salt diet (HSD) is associated with blood pressure regulation. Lymph nodes, located along lymphatic pathways, are not only important secondary lymphoid tissues for cancer metastasis, inflammation, and immune responses, but are also important for fluid homeostasis. Afferent lymphatics collect lymph from the pre-nodal area and efferent lymphatics drain lymph out of the lymph nodes. However, the difference in mechanical activity between afferent and efferent lymphatics and the effect of a HSD on these vessels have not been shown. Changes in mechanical activity of isolated afferent and efferent lymphatics in normal salt diet (NSD) and 4-week HSD mice in response to increases in intraluminal pressures from 3 to 7 cmH2O were measured using video-microscopy. The higher intramural pressure equivalently decreased pumping activity of afferent and efferent lymphatics in NSD mice. A HSD suppressed the amplitude, ejection fraction, and stroke volume of afferent lymphatics, leading to marked reductions in pumping activity. In contrast, the pumping activities of efferent lymphatics were resistant to a HSD and were preserved by enhancing the contraction frequency. A HSD differentially modulated the mechanical activity of afferent and efferent collecting lymphatics in murine iliac lymph nodes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7634
Nifedipine, ≥98% (HPLC), powder
C17H18N2O6
N114
Nifedipine
C17H18N2O6