PloS one
Defining the Pharmacodynamic Profile and Therapeutic Index of NHS-IL12 Immunocytokine in Dogs with Malignant Melanoma.
PMID 26091536
Abstract
Interleukin (IL)-12 is a pro-inflammatory cytokine that mediates T-helper type 1 responses and cytotoxic T-cell activation, contributing to its utility as anti-cancer agent. Systemic administration of IL-12 often results in unacceptable toxicity; therefore, strategies to direct delivery of IL-12 to tumors are under investigation. The objective of this study was to assist the preclinical development of NHS-IL12, an immunocytokine consisting of an antibody, which targets necrotic tumor regions, linked to IL-12. Specifically this study sought to evaluate the safety, serum pharmacokinetics, anti-tumor activity, and immune modulation of NHS-IL12 in dogs with naturally occurring cancers. A rapid dose-escalation study of NHS-IL12 administered subcutaneously to dogs with melanoma was conducted through the Comparative Oncology Trials Consortium (COTC). Eleven dogs were enrolled in four dose-escalation cohorts; thereafter, an additional seven dogs were treated at the defined tolerable dose of 0.8 mg/m2. The expanded cohort at this fixed dose (ten dogs in total) was accrued for further pharmacokinetics and pharmacodynamics assessment. NHS-IL12 levels, serum cytokine concentrations, and peripheral blood mononuclear cell characterization (post-treatment) and draining lymph node immune profiling, and tumor biopsies (pre- and post-treatment) were collected. Adverse events included thrombocytopenia, liver enzymopathies, fever, and vasculitis. Correlation between interferon (IFN)-γ induction, adverse events, and NHS-IL12 exposure (maximum concentration and area under the concentration-time curve) were dose-dependent. Serum IL-10 levels and intratumoral CD8+ populations increased after treatment. Partial responses, according to Response Evaluation Criteria in Solid Tumors (RECIST) criteria, were observed in two dogs treated with NHS-IL12 0.8 mg/m2 and 1.6 mg/m2. NHS-IL12 was administered safely to dogs with melanoma and both immunologic and clinical activity was observed. This study successfully defined a narrow therapeutic window for systemic delivery of NHS-IL12 via the subcutaneous route. Results will inform the design and implementation of first-in-human clinical trials of NHS-IL12 in cancer patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60339
Potassium bicarbonate, BioUltra, ≥99.5% (T)
CHKO3