PloS one
NLRP3 Inflammasome Is Expressed and Functional in Mouse Brain Microglia but Not in Astrocytes.
PMID 26091541
Abstract
Neuroinflammation is the local reaction of the brain to infection, trauma, toxic molecules or protein aggregates. The brain resident macrophages, microglia, are able to trigger an appropriate response involving secretion of cytokines and chemokines, resulting in the activation of astrocytes and recruitment of peripheral immune cells. IL-1β plays an important role in this response; yet its production and mode of action in the brain are not fully understood and its precise implication in neurodegenerative diseases needs further characterization. Our results indicate that the capacity to form a functional NLRP3 inflammasome and secretion of IL-1β is limited to the microglial compartment in the mouse brain. We were not able to observe IL-1β secretion from astrocytes, nor do they express all NLRP3 inflammasome components. Microglia were able to produce IL-1β in response to different classical inflammasome activators, such as ATP, Nigericin or Alum. Similarly, microglia secreted IL-18 and IL-1α, two other inflammasome-linked pro-inflammatory factors. Cell stimulation with α-synuclein, a neurodegenerative disease-related peptide, did not result in the release of active IL-1β by microglia, despite a weak pro-inflammatory effect. Amyloid-β peptides were able to activate the NLRP3 inflammasome in microglia and IL-1β secretion occurred in a P2X7 receptor-independent manner. Thus microglia-dependent inflammasome activation can play an important role in the brain and especially in neuroinflammatory conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS2287
Anti-ASC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503273
Anti-CASP1 (p20, Cleaved-Asn120) antibody produced in rabbit, affinity isolated antibody
HPA003056
Anti-CASP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503271
Anti-CASP1, C-Terminal antibody produced in rabbit, affinity isolated antibody
PRS3459
Anti-Caspase-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3463
Anti-Caspase-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV38110
Anti-HMGB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2701685
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701809
Anti-hmgb1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101049
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB4501401
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
AV54322
Anti-IL1A antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1406017
Anti-IL1B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV54324
Anti-IL1B antibody produced in rabbit, affinity isolated antibody
SAB1409150
Anti-PYCARD antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV37320
Anti-PYCARD antibody produced in rabbit, IgG fraction of antiserum
SAB1407378
Anti-PYCARD, (N-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
RAB0267
Human IL-18 BPa  ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0268
Human IL-18 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0543
Human IL18 / Interleukin-18 ELISA Kit, for serum, plasma, cell culture supernatants and urine
WH0000834M1
Monoclonal Anti-CASP1 antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
H9537
Monoclonal Anti-HMG-1 antibody produced in mouse, clone HAP46.5, purified from hybridoma cell culture
WH0003146M8
Monoclonal Anti-HMGB1 antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
SAB1403925
Monoclonal Anti-HMGB1 antibody produced in mouse, clone 1D5, purified immunoglobulin, buffered aqueous solution
WH0003552M1
Monoclonal Anti-IL1A antibody produced in mouse, clone 4B8, purified immunoglobulin, buffered aqueous solution
SAB1403967
Monoclonal Anti-IL1A, (C-terminal) antibody produced in mouse, clone 1F3, purified immunoglobulin, buffered aqueous solution
WH0003553M1
Monoclonal Anti-IL1B antibody produced in mouse, clone 2A8, purified immunoglobulin, buffered aqueous solution
SAB1403968
Monoclonal Anti-IL1B antibody produced in mouse, clone 2E8, purified immunoglobulin, buffered aqueous solution
RAB0810 Mouse Il18 / Interleukin-18 ELISA Kit, for serum, plasma and cell culture supernatants