EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Reduction of DNA damage induced by titanium dioxide nanoparticles through Nrf2 in vitro and in vivo.
PMID 26091733
Abstract
Titanium dioxide nanoparticles (Nano-TiO2) are widely used to additives in cosmetics, pharmaceutical, paints and foods. Recent studies have demonstrated that Nano-TiO2 induces DNA damage and increased the risk of cancer and the mechanism might relate with oxidative stress. The aim of this study was to evaluate the effects of Nuclear factor erythroid 2 (NF-E2)-related factor 2 (Nrf2), an anti-oxidative mediator, on DNA damage induced by Nano-TiO2. Wildtype, Nrf2 knockout (Nrf2(-/-)) and tert-butylhydroquinone (tBHQ) pre-treated HepG2 cells and mice were treated with Nano-TiO2. And then the oxidative stress and DNA damage were evaluated. Our data showed that DNA damage, reactive oxygen species (ROS) generation and MDA content in Nano-TiO2 exposed cells were significantly increased than those of control in dose dependent manners. Nrf2/ARE droved the downstream genes including NAD(P)H dehydrogenase [quinine] 1(NQO1), heme oxygenase 1 (HO-1) and glutamate-cysteine ligase catalytic subunit (GCLC) expression were significantly higher in wildtype HepG2 cells after Nano-TiO2 treatment. After treatment with Nano-TiO2, the DNA damages were significantly increased in Nrf(-/-) cells and mice whereas significantly decreased in tBHQ pre-treatment cells and mice, compared with the wildtype HepG2 cells and mice, respectively. Our results indicated that the acquired of Nrf2 leads to a decreased susceptibility to DNA damages induction by Nano-TiO2 and decreasing of risk of cancer which would provide a strategy for a more efficacious sensitization of against of Nano-TiO2 toxication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU082841 MISSION® esiRNA, esiRNA targeting mouse Nfe2l2 (esiRNA1)