EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Effects of Xylo-Oligosaccharides on Broiler Chicken Performance and Microbiota.
PMID 26092452
Abstract
In broiler chickens, feed additives, including prebiotics, are widely used to improve gut health and to stimulate performance. Xylo-oligosaccharides (XOS) are hydrolytic degradation products of arabinoxylans that can be fermented by the gut microbiota. In the current study, we aimed to analyze the prebiotic properties of XOS when added to the broiler diet. Administration of XOS to chickens, in addition to a wheat-rye-based diet, significantly improved the feed conversion ratio. XOS significantly increased villus length in the ileum. It also significantly increased numbers of lactobacilli in the colon and Clostridium cluster XIVa in the ceca. Moreover, the number of gene copies encoding the key bacterial enzyme for butyrate production, butyryl-coenzyme A (butyryl-CoA):acetate CoA transferase, was significantly increased in the ceca of chickens administered XOS. In this group of chickens, at the species level, Lactobacillus crispatus and Anaerostipes butyraticus were significantly increased in abundance in the colon and cecum, respectively. In vitro fermentation of XOS revealed cross-feeding between L. crispatus and A. butyraticus. Lactate, produced by L. crispatus during XOS fermentation, was utilized by the butyrate-producing Anaerostipes species. These data show the beneficial effects of XOS on broiler performance when added to the feed, which potentially can be explained by stimulation of butyrate-producing bacteria through cross-feeding of lactate and subsequent effects of butyrate on gastrointestinal function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
309435
3-Methyl-1-butanol, anhydrous, ≥99%
C5H12O
I9392
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
C5H12O
59085
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
C5H12O
W360600
D-(+)-Xylose, ≥99%
C5H10O5
X1500
D-(+)-Xylose, ≥99%
C5H10O5
X3877
D-(+)-Xylose, BioXtra, ≥99%
C5H10O5
95729
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C5H10O5
ARK2188
Hexadecyltrimethylammonium bromide, PharmaGrade, USP/NF, Manufactured under appropriate controls for pharma or biopharmaceutical production.
C19H42BrN
H9151
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
C19H42BrN
52365
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
C19H42BrN
H6269
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
C19H42BrN
H5882
Hexadecyltrimethylammonium bromide, ≥98%
C19H42BrN
W205702
Isoamyl alcohol, ≥98%, FG
C5H12O
W205710
Isoamyl alcohol, natural, ≥98%, FG
C5H12O
L1376
Linoleic acid, ≥99%
C18H32O2
L1012
Linoleic acid, liquid, BioReagent, suitable for cell culture
C18H32O2
L1268
Linoleic acid, ≥93% (GC)
C18H32O2
W338001
Linoleic acid, ≥95%
C18H32O2
62240
Linoleic acid, technical, 60-74% (GC)
C18H32O2
P2222
Potassium phosphate dibasic, meets USP testing specifications
HK2O4P
450200
Potassium phosphate dibasic, 99.95% trace metals basis
HK2O4P
P8281
Potassium phosphate dibasic, reagent grade, ≥98.0%
HK2O4P
60353
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
HK2O4P
P8584
Potassium phosphate dibasic solution, 1.0 M
HK2O4P