EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Comparative analysis of antimicrobial resistance and genetic diversity of Campylobacter from broilers slaughtered in Poland.
PMID 26092707
Abstract
In the current study, the relationship of Campylobacter jejuni and Campylobacter coli strains isolated at slaughter was investigated using comparative analysis of antimicrobial resistance (AMR), virulence gene (VG) and PFGE profiling. A total of 254 Campylobacter isolates from poultry caeca and corresponding carcasses, including 139 C. jejuni and 115 C. coli strains were tested. The most prevalent resistance profiles observed in C. jejuni were ciprofloxacin, nalidixic acid and tetracycline (46 out of 139, 33.1% isolates) as well as ciprofloxacin, nalidixic acid, tetracycline and streptomycin among C. coli strains (34 out of 115, 29.6%). Multi-resistance was found more frequently among C. coli than C. jejuni (P<0.05). The presence of 11 virulence genes exhibited 19 different VG profiles in Campylobacter isolates tested. All Campylobacter strains were classified into 154 different PFGE types. Among them, 56 profiles (28 C. jejuni and 28 C. coli) were common for at least two isolates including 9 clusters covering from 4 to 9 strains. Campylobacter composite types generated by a combination of 154 PFGE types, 10 AMR profiles and 19 VG patterns divided 178 distinct types with 95% similarity. The majority of the composite profiles (76 for C. jejuni and 58 for C. coli; 75.3% in total) included only one bacterial isolate. Furthermore, 11 pairs of C. jejuni and 12 pairs of C. coli from caeca and the corresponding carcasses isolated from the same places possessed the identical PFGE, AMR and VG patterns. This study demonstrated that C. jejuni and C. coli isolated from poultry in Poland showed to have a high genetic diversity and a weak clonal population structure. However, the composite analysis revealed a strong evidence for cross-contamination of chicken carcasses during the slaughter process. Additionally, our results confirm that Campylobacter may easily contaminate poultry carcasses at slaughter process and spread around country. More than half of Campylobacter strains tested (50.4%) were resistant to at least two classes of antimicrobials, i.e. quinolones and tetracyclines, which may cause a public health risk.