EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
C3N4-sensitized TiO2 nanotube arrays with enhanced visible-light photoelectrochemical performance.
PMID 26095326
Abstract
A C3N4-sensitized TiO2 nanotube array-based photoanode was designed and fabricated via the in situ growth of C3N4 on the surface of TiO2 nanotubes. It shows stable and significantly improved PEC activity for hydrogen generation under visible light irradiation with a hydrogen production rate of about 19.1 μmol h(-1) (a Faradaic efficiency of nearly 100%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN