EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
Chemical and organoleptic characteristics of tomato purée enriched with lyophilized tomato pomace.
PMID 26095330
Abstract
Epidemiological studies have proved that tomato consumption is associated with a lower risk of developing several diseases (for example, certain types of cancers, cardiovascular diseases, macular degeneration, age-related eye disease). Many micronutrients and bioactive compounds are mainly present in peel and seeds and are lost during the processing into sauce, purée, paste and juice. The addition of lyophilized and powdered tomato pomace enhances the properties of purée. In this paper we report the chemical and physicochemical characterization of a purée enriched with 2% dry pomace. Comparison of the analytical data of starting purée with the enriched purée showed a significant increase of all micronutrients, without the taste and appearance being compromised or altered negatively. The product obtained is an example of a functional food rich in health-promoting phytochemicals, with the significant aspect of recovering a waste fraction of the tomato processing that would normally be disposed of in landfill, with associated costs and environmental impact.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M47949
2-Methylheptane, 98%
C8H18
C5157
Campesterol, ~65%
C28H48O
14681
Zeaxanthin, analytical standard
C40H56O2