EMAIL THIS PAGE TO A FRIEND
Reproductive biomedicine online
Deliveries from trophectoderm biopsied, fresh and vitrified blastocysts derived from polar body biopsied, vitrified oocytes.
PMID 26096028
Abstract
This longitudinal study reports preliminary findings of six patients who underwent first polar body biopsy followed by oocyte vitrification. All oocytes were warmed, inseminated by intracytoplasmic sperm injection and cultured to blastocyst. All suitable blastocysts underwent trophectoderm biopsy for aneuploidy screening, and supernumerary blastocysts were vitrified. Euploid blastocysts were transferred either fresh or in a subsequent programmed cycle. Of the 91 metaphase II oocytes, 30 had euploid first polar bodies. Development to blastocyst was more likely in oocytes with a euploid first polar body (66.7% versus 24.6%; P < 0.001). Nineteen euploid blastocysts were produced: 10 from oocytes with a euploid first polar body and nine from oocytes with an aneuploid first polar body. Five out of six patients (83%) had a live birth or ongoing pregnancy at the time of analysis. Eleven euploid blastocysts have been transferred and seven implanted (64%). Although the chromosomal status of the first polar body was poorly predictive of embryonic ploidy, an association was found between chromosomal status of the first polar body and development to blastocyst. Further study is required to characterize these relationships, but proof of concept is provided that twice biopsied, twice cryopreserved oocytes and embryos can lead to viable pregnancies.