PloS one
Mest but Not MiR-335 Affects Skeletal Muscle Growth and Regeneration.
PMID 26098312
Abstract
When skeletal muscle fibers are injured, they regenerate and grow until their sizes are adjusted to surrounding muscle fibers and other relevant organs. In this study, we examined whether Mest, one of paternally expressed imprinted genes that regulates body size during development, and miR-335 located in the second intron of the Mest gene play roles in muscle regeneration. We generated miR-335-deficient mice, and found that miR-335 is a paternally expressed imprinted microRNA. Although both Mest and miR-335 are highly expressed during muscle development and regeneration, only Mest+/- (maternal/paternal) mice show retardation of body growth. In addition to reduced body weight in Mest+/-; DMD-null mice, decreased muscle growth was observed in Mest+/- mice during cardiotoxin-induced regeneration, suggesting roles of Mest in muscle regeneration. Moreover, expressions of H19 and Igf2r, maternally expressed imprinted genes were affected in tibialis anterior muscle of Mest+/-; DMD-null mice compared to DMD-null mice. Thus, Mest likely mediates muscle regeneration through regulation of imprinted gene networks in skeletal muscle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15-2230
2-Methylbutane, SAJ first grade, ≥99.0%
C5H12
AV32393
Anti-PAX7 (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV32742
Anti-PAX7 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1412356
ANTI-PAX7 antibody produced in mouse, clone 1G11, purified immunoglobulin, buffered aqueous solution
SAB1412376
ANTI-PAX7 antibody produced in mouse, clone 3H1, purified immunoglobulin, buffered aqueous solution
SAB1412455
ANTI-PAX7 antibody produced in mouse, clone 4F8, purified immunoglobulin, buffered aqueous solution
SAB1412599
ANTI-PAX7 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB1412600
ANTI-PAX7 antibody produced in mouse, clone 3F10, purified immunoglobulin, buffered aqueous solution
SAB1412601
ANTI-PAX7 antibody produced in mouse, clone 4F5, purified immunoglobulin, buffered aqueous solution
SAB2101730
Anti-PAX7 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2103961
Anti-PAX7 antibody produced in rabbit, affinity isolated antibody
WH0005081M3
Monoclonal Anti-PAX7 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
SAB1404168
Monoclonal Anti-PAX7, (C-terminal) antibody produced in mouse, clone 1E12, purified immunoglobulin, buffered aqueous solution