EMAIL THIS PAGE TO A FRIEND
Oncotarget
The LPA1/ZEB1/miR-21-activation pathway regulates metastasis in basal breast cancer.
PMID 26098771
Abstract
Lysophosphatidic acid (LPA) is a bioactive lipid promoting cancer metastasis. LPA activates a series of six G protein-coupled receptors (LPA1-6). While blockage of LPA1in vivo inhibits breast carcinoma metastasis, down-stream genes mediating LPA-induced metastasis have not been yet identified. Herein we showed by analyzing publicly available expression data from 1488 human primary breast tumors that the gene encoding the transcription factor ZEB1 was the most correlated with LPAR1 encoding LPA1. This correlation was most prominent in basal primary breast carcinomas and restricted to cell lines of basal subtypes. Functional experiments in three different basal cell lines revealed that LPA-induced ZEB1 expression was regulated by the LPA1/Phosphatidylinositol-3-Kinase (Pi3K) axis. DNA microarray and real-time PCR analyses further demonstrated that LPA up-regulated the oncomiR miR-21 through an LPA1/Pi3K/ZEB1-dependent mechanism. Strikingly, treatment with a mirVana miR-21 inhibitor, or silencing LPA1 or ZEB1 completely blocked LPA-induced cell migration in vitro, invasion and tumor cell bone colonization in vivo, which can be restored with a mirVana miR-21 mimic. Finally, high LPAR1 expression in basal breast tumors predicted worse lung-metastasis-free survival. Collectively, our results elucidate a new molecular pathway driving LPA-induced metastasis, thus underscoring the therapeutic potential of targeting LPA1 in patients with basal breast carcinomas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV100657
Anti-AREB6 antibody produced in rabbit, affinity isolated antibody
GW21144 Anti-c-Fos antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1306144
ANTI-C-FOS(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2100832
Anti-FOS (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100833
Anti-FOS (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2502116
Anti-FOS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104185
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500995
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500993
Anti-FOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500996
Anti-FOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300327
Anti-STAT3 (Ab-727) antibody produced in rabbit, affinity isolated antibody
SAB2500993
Anti-STAT3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406487
Anti-STAT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2102317
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104461
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104912
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB4502871
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502872
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502873
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA027524
Anti-ZEB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102759
Anti-ZEB1 antibody produced in rabbit, affinity isolated antibody
SAB3500514
Anti-ZEB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
300306
Methylcyclohexane, anhydrous, ≥99%
C7H14
EHU097831 MISSION® esiRNA, esiRNA human LPAR1 (esiRNA1)
EMU054251 MISSION® esiRNA, esiRNA targeting mouse Lpar1 (esiRNA1)
SAB5300463
Monoclonal Anti-FOS antibody produced in mouse, clone 2G2, ascites fluid
SAB1402201
Monoclonal Anti-FOS antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
WH0006774M2
Monoclonal Anti-STAT3 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
AMAB90777
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0492, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90776
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0490, purified immunoglobulin, buffered aqueous glycerol solution