EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Collapse of Particle-Laden Interfaces under Compression: Buckling vs Particle Expulsion.
PMID 26099031
Abstract
Colloidal particles can bind to fluid interfaces with a capillary energy that is thousands of times the thermal energy. This phenomenon offers an effective route to emulsion and foam stabilization where the stability is influenced by the phase behavior of the particle-laden interface under deformation. Despite the vast interest in particle-laden interfaces, the key factors that determine the collapse of such an interface under compression have remained relatively unexplored. In this study, we illustrate the significance of the particle surface wettability and presence of electrolyte in the subphase on interparticle interactions at the interface and the resulting collapse mode. Various collapse mechanisms including buckling, particle expulsion, and multilayer formation are reported and interpreted in terms of particle-particle and particle-interface interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
440272
Dichlorodimethylsilane, ≥99.5%
C2H6Cl2Si
40140
Dichlorodimethylsilane, ≥98.5% (GC)
C2H6Cl2Si