EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
RNA-Binding Protein Musashi1 Is a Central Regulator of Adhesion Pathways in Glioblastoma.
PMID 26100017
Abstract
The conserved RNA-binding protein Musashi1 (MSI1) has emerged as a key oncogenic factor in numerous solid tumors, including glioblastoma. However, its mechanism of action has not yet been established comprehensively. To identify its target genes comprehensively and determine the main routes by which it influences glioblastoma phenotypes, we conducted individual-nucleotide resolution cross-linking and immunoprecipitation (iCLIP) experiments. We confirmed that MSI1 has a preference for UAG sequences contained in a particular structural context, especially in 3' untranslated regions. Although numerous binding sites were also identified in intronic sequences, our RNA transcriptome sequencing analysis does not favor the idea that MSI1 is a major regulator of splicing in glioblastoma cells. MSI1 target mRNAs encode proteins that function in multiple pathways of cell proliferation and cell adhesion. Since these associations indicate potentially new roles for MSI1, we investigated its impact on glioblastoma cell adhesion, morphology, migration, and invasion. These processes are known to underpin the spread and relapse of glioblastoma, in contrast to other tumors where metastasis is the main driver of recurrence and progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21062 Anti-CD221 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300480
Anti-EGFR (Ab-1197) antibody produced in rabbit, affinity isolated antibody
SAB4300453
Anti-EGFR (Ab-678) antibody produced in rabbit, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
SAB4300481
Anti-EGFR (Ab-869) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018530
Anti-EGFR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500720
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500713
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500714
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500715
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500716
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500717
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500718
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500719
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500721
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500722
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500723
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4300359
Anti-IGF1R (Ab-1161) antibody produced in rabbit, affinity isolated antibody
SAB4300360
Anti-IGF1R (Ab-1165/1166) antibody produced in rabbit, affinity isolated antibody