EMAIL THIS PAGE TO A FRIEND
SpringerPlus
Unmanipulated native fat exposed to high-energy diet, but not autologous grafted fat by itself, may lead to overexpression of Ki67 and PAI-1.
PMID 26101731
Abstract
Although its unclear oncological risk, which led to more than 20xa0years of prohibition of its use, fat grafting to the breast is widely used nowadays even for aesthetic purposes. Thus, we proposed an experimental model in rats to analyze the inflammatory activity, cellular proliferation and levels of Plasminogen Activator Inhibitor (PAI-1) in grafted fat, and in native fat exposed to high-energy diet in order to study the oncological potential of fat tissue. Samples of grafted fat of rats on regular-energy diet were compared with paired samples of native fat from the same rat on regular-energy diet and on high-energy diet in a different time. Analysis involved microscopic comparisons using hematoxylin-eosin staining, immunohistochemistry with anti-CD68-labelled macrophages, and gene expression of Ki-67 and PAI-1. Hematoxylin-eosin staining analyses did not find any atypical cellular infiltration or unusual tissue types in the samples of grafted fat. The inflammatory status, assessed through immunohistochemical identification of CD68-labelled macrophages, was similar among samples of native fat and grafted fat of rat on regular-energy diet and of native fat of rats on high-energy diet. Real-time PCR revealed that high-energy diet, but not fat grafting, leads to proliferative status on adipose tissue (overexpression of ki-67, pxa0=xa00.046) and raised its PAI-1 levels, pxa0