EMAIL THIS PAGE TO A FRIEND
BioMed research international
Antiherpes Activity and Skin/Mucosa Distribution of Flavonoids from Achyrocline satureioides Extract Incorporated into Topical Nanoemulsions.
PMID 26101767
Abstract
This study investigated the inhibitory effects of Achyrocline satureioides extract (ASE) incorporated into a topical nanoemulsion on Herpes Simplex Virus type 1 (HSV-1/KOS strain) replication, as well as the distribution of the main ASE flavonoids (quercetin, luteolin, and 3-O-methylquercetin) in porcine skin and mucosa. The ASE-loaded nanoemulsion showed more pronounced effects against HSV-1 replication when compared to the ASE or pure quercetin, as determined by the viral plaque number reduction assay. All flavonoids were detected in the skin epidermis (2.2 µg/cm(2)) and the mucosa upper layers (3.0 µg/cm(2)) from ASE-loaded nanoemulsion until 8 h after topical application. A higher amount of flavonoids was detected when these tissues were impaired, especially in deeper mucosa layers (up to 7-fold). Flavonoids were detected in the receptor fluid only when the mucosa was injured. Such results were supported by confocal microscopy images. Overall, these findings suggest that the tested ASE-loaded nanoemulsion has potential to be used topically for herpes infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3251
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
C29H50O2
T3634
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
C29H50O2
T1539
(+)-α-Tocopherol, Type VI, from vegetable oil, neat (liquid, ≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
C29H50O2
90081
3-O-Methylquercetin, ≥97% (HPLC)
C16H12O7
258024
α-Tocopherol, ≥95.5%
C29H50O2
L9283
Luteolin, ≥98% (TLC), powder
C15H10O6
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
Q4951
Quercetin, ≥95% (HPLC), solid
C15H10O7