EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Glycine transporter GlyT1, but not GlyT2, is expressed in rat dorsal root ganglion--Possible implications for neuropathic pain.
PMID 26101830
Abstract
Glycinergic inhibitory neurotransmission plays a pivotal role in the development of neuropathic pain. The glycine concentration in the synaptic cleft is controlled by the glycine transporters GlyT1 and GlyT2. GlyT1 is expressed throughout the central nervous system, while GlyT2 is exclusively located in glycinergic neurons. Aim of the present study was to investigate whether GlyTs are also expressed in the peripheral sensory nervous system and whether their expression is modulated in experimental neuropathic pain. Neuropathic pain was induced in male Wistar rats by Chronic Constriction Injury (CCI) and verified by assessment of mechanical allodynia (von Frey method). Expression patterns of GlyTs and the glycine binding subunit NR1 of the N-methyl-d-aspartate (NMDA) receptor in the spinal cord and dorsal root ganglia (DRG) were analyzed by Western blot analysis, PCR and immunohistochemistry. While both GlyT1 and GlyT2 were detected in the spinal cord, only GlyT1, but not GlyT2, was detected in DRG. Immunofluorescence revealed a strictly neuronal localization of GlyT1 and a co-localization of GlyT1 and NR1 in DRG. Compared to sham procedure, spinal cord and DRG expression of GlyT1 was not altered and NR1 was unchanged in DRG 12 days after CCI. GlyT1, but not GlyT2, is expressed in the peripheral sensory nervous system. The co-expression of GlyT1 and NMDA receptors in DRG suggests that GlyT1 regulates glycine concentration at the glycine binding site of the NMDA receptor. Differential regulation of GlyT1 expression in the spinal cord or DRG, however, does not seem to be associated with the development of neuropathic pain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100019
Anti-ACAA2 antibody produced in rabbit, affinity isolated antibody
HPA042303
Anti-ACAA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503585
Anti-CD5, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300405
Anti-GRIN1 (Ab-896) antibody produced in rabbit, affinity isolated antibody
SAB4300538
Anti-GRIN1 (Ab-897) antibody produced in rabbit, affinity isolated antibody
HPA007406
Anti-IL1RL1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101150
Anti-IL1RL1 antibody produced in rabbit, affinity isolated antibody
HPA007917
Anti-IL1RL1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024504
Anti-NDOR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410533
Anti-NDOR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501301
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501302
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501303
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2105530
Anti-SLC25A4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0010449M1
Monoclonal Anti-ACAA2 antibody produced in mouse, clone 5C4, purified immunoglobulin, buffered aqueous solution
SAB1404828
Monoclonal Anti-ACAA2 antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
WH0027158M1
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 3A11, purified immunoglobulin, buffered aqueous solution
SAB1405033
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 3A11, ascites fluid
SAB4100604
Monoclonal Anti-NDOR1 antibody produced in mouse, clone NDOR1-A590, culture supernatant
SAB1409118
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 5A7, purified immunoglobulin, buffered aqueous solution