EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Size- and shape-dependent peroxidase-like catalytic activity of MnFe2O4 Nanoparticles and their applications in highly efficient colorimetric detection of target cancer cells.
PMID 26102593
Abstract
The catalytic activity of nanocrystal catalysts depends strongly on their chemical composition, size, and shape. Herein, we report four different sizes and shapes of MnFe2O4 nanoparticles (NPs) prepared by a hydrothermal procedure. In addition, the size- and shape-dependent peroxidase-like activity of these NPs was first explored using 3,3',5,5'-tetramethyl-benzidine and H2O2 as peroxidase substrates. The results showed that the peroxidase-like activities of the MnFe2O4 NPs were size- and shape-dependent and followed the order of 4 nm (spherical) > 18 nm (plate-like) > 27 nm (near-cubic) > 16 nm (spherical); this order was closely related to their surface-to-volume ratio and atom arrangements. Such an investigation is of great significance for peroxidase nanomimetics with enhanced activity and utilization. Furthermore, folic acid (FA)-conjugated MnFe2O4 NPs allow the detection of folate receptor-rich cancer cells. Such investigation can be widely utilized for the identification of important target molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245763
Manganese(II) acetylacetonate
C10H14MnO4