EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Solid-State (17)O NMR of Oxygen-Nitrogen Singly Bonded Compounds: Hydroxylammonium Chloride and Sodium Trioxodinitrate (Angeli's Salt).
PMID 26107984
Abstract
We report a solid-state NMR study of (17)O-labeled hydroxylammonium chloride ([H(17)O-NH3]Cl) and sodium trioxodinitrate monohydrate (Na2[(17)ONNO2]·H2O, Angeli's salt). The common feature in these two compounds is that they both contain oxygen atoms that are singly bonded to nitrogen. For this class of oxygen-containing functional groups, there is very limited solid-state (17)O NMR data in the literature. In this work, we experimentally measured the (17)O chemical shift and quadrupolar coupling tensors. With the aid of plane-wave DFT computation, the (17)O NMR tensor orientations were determined in the molecular frame of reference. We found that the characteristic feature of an O-N single bond is that the (17)O nucleus exhibits a large quadrupolar coupling constant (13-15 MHz) but a rather small chemical shift anisotropy (100-250 ppm), in sharp contrast with the nitroso (O═N) functional group for which both quantities are very large (e.g., 16 MHz and 3000 ppm, respectively).