EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Effects of rhodiola crenulata on mice hearts under severe sleep apnea.
PMID 26108210
Abstract
The goal of this study is to determine if Rhodiola Crenulata (RC) has protective effects on mice hearts with severe sleep apnea model. Sixty-four C57BL/6 J mice 5-6 months old were distributed into 4 groups i.e. Control group (21% O2, 24 h per day, 8 weeks, n=16); Hypoxia group (Hypoxia: 7% O2 60 s, 20% O2 alternating 60 s, 8 h per day, 8 weeks, n=16); Hypoxia+90RC and Hypoxia+270RC group (Hypoxia for 1st 4 weeks and hypoxia pretreated 90 mg/Kg and 270 mg/Kg Rhodiola Crenulata by oral gavage per day for 2nd 4 weeks, each n=16). Excised hearts from 4 groups of mice were analyzed for heart weight index changes using H&E staining, TUNEL-positive assays and Western Blotting protein. Cardiac widely dispersed TUNEL-positive apoptotic cells in mice hearts were less in Hypoxia+RC90 and Hypoxia+RC270 than those in Hypoxia. Compared with Hypoxia, the protein levels of Fas ligand, Fas death receptors, Fas-Associated Death Domain (FADD), activated caspase 8, and activated caspase 3 (Fas dependent apoptotic pathways) were decreased in Hypoxia+RC90, Hypoxia+RC270. The protein levels of Bad, Bax, t-Bid, activated caspase 9, activated caspase 3 (mitochondria dependent apoptotic pathway) were less in Hypoxia+RC90, Hypoxia+RC270 than those in hypoxia. The protein levels of Bcl2, Bcl-xL, p-Bad (Bcl2-realted anti-apoptotic pathway) and VEGF, p-PI3k, p-AKT (VEGF-related pro-survival pathway) were higher in Hypoxia+RC90, Hypoxia+RC270 than those in hypoxia. Our findings suggest that Rhodiola Crenulata have protective effects on chronic intermittent hypoxia-induced cardiac widely dispersed apoptosis via Fas-dependent and mitochondria-dependent apoptotic and VEGF-related pro-survival pathway.