EMAIL THIS PAGE TO A FRIEND
Nature communications
The sorting protein PACS-2 promotes ErbB signalling by regulating recycling of the metalloproteinase ADAM17.
PMID 26108729
Abstract
The metalloproteinase ADAM17 activates ErbB signalling by releasing ligands from the cell surface, a key step underlying epithelial development, growth and tumour progression. However, mechanisms acutely controlling ADAM17 cell-surface availability to modulate the extent of ErbB ligand release are poorly understood. Here, through a functional genome-wide siRNA screen, we identify the sorting protein PACS-2 as a regulator of ADAM17 trafficking and ErbB signalling. PACS-2 loss reduces ADAM17 cell-surface levels and ADAM17-dependent ErbB ligand shedding, without apparent effects on related proteases. PACS-2 co-localizes with ADAM17 on early endosomes and PACS-2 knockdown decreases the recycling and stability of internalized ADAM17. Hence, PACS-2 sustains ADAM17 cell-surface activity by diverting ADAM17 away from degradative pathways. Interestingly, Pacs2-deficient mice display significantly reduced levels of phosphorylated EGFR and intestinal proliferation. We suggest that this mechanism controlling ADAM17 cell-surface availability and EGFR signalling may play a role in intestinal homeostasis, with potential implications for cancer biology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33510
1,10-Phenanthroline monohydrate, ACS reagent, puriss. p.a., ≥99.5% (calc. to the dried substance), for redox titration
C12H8N2 · H2O
320056
1,10-Phenanthroline monohydrate, ACS reagent, 99%
C12H8N2 · H2O
P9375
1,10-Phenanthroline monohydrate, reagent grade
C12H8N2 · H2O
N9389
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N2765
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N2640
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N4645
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
C6H4NNa2O6P · 6H2O
71768
4-Nitrophenyl phosphate disodium salt hexahydrate, for enzyme immunoassay, ≥99.0% (enzymatic)
C6H4NNa2O6P · 6H2O
SML0041
Batimastat, ≥98% (HPLC)
C23H31N3O4S2
EMU037311 MISSION® esiRNA, esiRNA targeting mouse Adam17 (esiRNA1)
EHU075381 MISSION® esiRNA, esiRNA human ADAM17 (esiRNA1)
EHU136321 MISSION® esiRNA, esiRNA human PACS2 (esiRNA1)
EMU183501 MISSION® esiRNA, esiRNA targeting mouse Pacs2
P4744
Phosphatase substrate, powder
C6H4NNa2O6P · 6H2O
S0942
Phosphatase substrate, 5 mg tablets
C6H4NNa2O6P · 6H2O
P5994
Phosphatase substrate, 40 mg tablets
C6H4NNa2O6P · 6H2O
P5744
Phosphatase substrate, 40 mg capsules
C6H4NNa2O6P · 6H2O
P5869
Phosphatase substrate, 100 mg capsules
C6H4NNa2O6P · 6H2O
SRE0026
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
C6H4NNa2O6P · 6H2O