EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Disturbances in Bone Largely Predict Aortic Calcification in an Alternative Rat Model Developed to Study Both Vascular and Bone Pathology in Chronic Kidney Disease.
PMID 26108730
Abstract
Because current rat models used to study chronic kidney disease (CKD)-related vascular calcification show consistent but excessive vascular calcification and chaotic, immeasurable, bone mineralization due to excessive bone turnover, they are not suited to study the bone-vascular axis in one and the same animal. Because vascular calcification and bone mineralization are closely related to each other, an animal model in which both pathologies can be studied concomitantly is highly needed. CKD-related vascular calcification in rats was induced by a 0.25% adenine/low vitamin K diet. To follow vascular calcification and bone pathology over time, rats were killed at weeks 4, 8, 10, 11, and 12. Both static and dynamic bone parameters were measured. Vascular calcification was quantified by histomorphometry and measurement of the arterial calcium content. Stable, severe CKD was induced along with hyperphosphatemia, hypocalcemia as well as increased serum PTH and FGF23. Calcification in the aorta and peripheral arteries was present from week 8 of CKD onward. Four and 8 weeks after CKD, static and dynamic bone parameters were measurable in all animals, thereby presenting typical features of hyperparathyroid bone disease. Multiple regression analysis showed that the eroded perimeter and mineral apposition rate in the bone were strong predictors for aortic calcification. This rat model presents a stable CKD, moderate vascular calcification, and quantifiable bone pathology after 8 weeks of CKD and is the first model that lends itself to study these main complications simultaneously in CKD in mechanistic and intervention studies.