EMAIL THIS PAGE TO A FRIEND
Genes & development
DNA methylation restrains transposons from adopting a chromatin signature permissive for meiotic recombination.
PMID 26109049
Abstract
DNA methylation is essential for protecting the mammalian germline against transposons. When DNA methylation-based transposon control is defective, meiotic chromosome pairing is consistently impaired during spermatogenesis: How and why meiosis is vulnerable to transposon activity is unknown. Using two DNA methylation-deficient backgrounds, the Dnmt3L and Miwi2 mutant mice, we reveal that DNA methylation is largely dispensable for silencing transposons before meiosis onset. After this, it becomes crucial to back up to a developmentally programmed H3K9me2 loss. Massive retrotransposition does not occur following transposon derepression, but the meiotic chromatin landscape is profoundly affected. Indeed, H3K4me3 marks gained over transcriptionally active transposons correlate with formation of SPO11-dependent double-strand breaks and recruitment of the DMC1 repair enzyme in Dnmt3L(-/-) meiotic cells, whereas these features are normally exclusive to meiotic recombination hot spots. Here, we demonstrate that DNA methylation restrains transposons from adopting chromatin characteristics amenable to meiotic recombination, which we propose prevents the occurrence of erratic chromosomal events.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100532
Anti-DBI antibody produced in rabbit, affinity isolated antibody
SAB1410285
Anti-DBI antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA001232
Anti-DMC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701985
Anti-DMC1 antibody produced in rabbit
HPA023095
Anti-HID1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA061260
Anti-HPS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C7657
Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
D5025
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
D4527
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
D4513
Deoxyribonuclease I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
D4263
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
DN25
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
D7291 Deoxyribonuclease I RNase-free solution from bovine pancreas
SAB4100149
Monoclonal Anti-C17orf28 antibody produced in mouse, clone C17orf28-A117, culture supernatant
WH0011144M7
Monoclonal Anti-DMC1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
SAB1404906
Monoclonal Anti-DMC1, (C-terminal) antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution
SAB5300087 Monoclonal Anti-HPS1 antibody produced in mouse, clone 5G12G2, ascites fluid
T6763 Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)