EMAIL THIS PAGE TO A FRIEND
Genes & development
Homeostatic control of Hippo signaling activity revealed by an endogenous activating mutation in YAP.
PMID 26109051
Abstract
The Hippo signaling pathway converges on YAP to regulate growth, differentiation, and regeneration. Previous studies with overexpressed proteins have shown that YAP is phosphorylated by its upstream kinase, Lats1/2, on multiple sites, including an evolutionarily conserved 14-3-3-binding site whose phosphorylation is believed to inhibit YAP by excluding it from the nucleus. Indeed, nuclear localization of YAP or decreased YAP phosphorylation at this site (S168 in Drosophila, S127 in humans, and S112 in mice) is widely used in current literature as a surrogate of YAP activation even though the physiological importance of this phosphorylation event in regulating endogenous YAP activity has not been defined. Here we address this question by introducing a Yap(S112A) knock-in mutation in the endogenous Yap locus. The Yap(S112A) mice are surprisingly normal despite nuclear localization of the mutant YAP protein in vivo and profound defects in cytoplasmic translocation in vitro. Interestingly, the mutant Yap(S112A) mice show a compensatory decrease in YAP protein levels due to increased phosphorylation at a mammalian-specific phosphodegron site on YAP. These findings reveal a robust homeostatic mechanism that maintains physiological levels of YAP activity and caution against the assumptive use of YAP localization alone as a surrogate of YAP activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
685720
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
C30H27N3
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003097
Anti-NF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
H0412 Anti-phospho-Histone H3 (pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503877
Anti-phospho-Histone H3 (pSer28) antibody produced in rabbit, affinity isolated antibody
SAB4503876
Anti-phospho-Histone H3 (pThr3) antibody produced in rabbit, affinity isolated antibody
SAB4504068
Anti-phospho-YAP (pSer127) antibody produced in rabbit, affinity isolated antibody
SAB1305603 ANTI-PHOSPHO-YAP(S127) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Y4645
Anti-phospho-YAP1 (pTyr357) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1305751
ANTI-YAP (CENTER S127) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500094
Anti-YAP antibody produced in rabbit, affinity isolated antibody
SAB4500095
Anti-YAP antibody produced in rabbit, affinity isolated antibody
Y4770
Anti-YAP1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2501938
Anti-YAP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV50530
Anti-YAP1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA006986
Anti-YY1AP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105760
Anti-YY1AP1 antibody produced in rabbit, affinity isolated antibody
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
H9908
Monoclonal Anti-phospho-Histone H3 (pSer28) antibody produced in rat, ~0.5 mg/mL, clone HTA28, purified immunoglobulin, buffered aqueous solution
Y0771 Monoclonal Anti-phospho-YAP1 (pTyr357) antibody produced in mouse, ~1.0 mg/mL, clone PYP-76, purified immunoglobulin, buffered aqueous solution
WH0010413M1
Monoclonal Anti-YAP1 antibody produced in mouse, clone 2F12, purified immunoglobulin, buffered aqueous solution
SAB1404823
Monoclonal Anti-YAP1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
SAB1402596
Monoclonal Anti-YY1AP1 antibody produced in mouse, clone 2G7, purified immunoglobulin, buffered aqueous solution