EMAIL THIS PAGE TO A FRIEND
Blood
RAP-011 improves erythropoiesis in zebrafish model of Diamond-Blackfan anemia through antagonizing lefty1.
PMID 26109203
Abstract
Diamond-Blackfan Anemia (DBA) is a bone marrow failure disorder characterized by low red blood cell count. Mutations in ribosomal protein genes have been identified in approximately half of all DBA cases. Corticosteriod therapy and bone marrow transplantation are common treatment options for patients; however, significant risks and complications are associated with these treatment options. Therefore, novel therapeutic approaches are needed for treating DBA. Sotatercept (ACE-011, and its murine ortholog RAP-011) acts as an activin receptor type IIA ligand trap, increasing hemoglobin and hematocrit in pharmacologic models, in healthy volunteers, and in patients with β-thalassemia, by expanding late-stage erythroblasts through a mechanism distinct from erythropoietin. Here, we evaluated the effects of RAP-011 in zebrafish models of RPL11 ribosome deficiency. Treatment with RAP-011 dramatically restored hemoglobin levels caused by ribosome stress. In zebrafish embryos, RAP-011 likely stimulates erythropoietic activity by sequestering lefty1 from erythroid cells. These findings identify lefty1 as a signaling component in the development of erythroid cells and rationalize the use of sotatercept in DBA patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9143
o-Dianisidine, peroxidase substrate
C14H16N2O2