EMAIL THIS PAGE TO A FRIEND
Pharmacognosy magazine
Assay method for quality control and stability studies of a new anti-diabetic and anti-dyslipidemic flavone (S002-853).
PMID 26109774
Abstract
Flavonoid-rich extract of the plant is long known for its anti-diabetic activities in traditional medicine. S002-853, a new flavone derivative synthesized by Central Drug Research Institute (CDRI) has been used for the present study. The present study aimed at development of an assay method for quality control (QC) and stability studies of a new anti-diabetic and anti-dyslipidemic agent CDRI compound S002-853. A validated high-performance liquid chromatography analysis method for S002-853 was developed for in process QC and stability studies. The separation was achieved on a RP-C18 (25 cm × 0.4 cm, 5 μm, Phenomenex) at 240 nm with flow rate of 1.0 ml/min. This method was applied successfully in establishing forced degradation and drug-excipient testing protocols as per International Conference on Harmonization guidelines. The result of estimation and stress testing studies indicated a high degree of selectivity of this method. S002-853 was most stable at pH 7 and under photolytic conditions. The temperature degradation pattern of S002-853 was found to follow the zero order degradation. The method described is easy and simple hence can be easily reproduced. This method can be very useful for bulk manufacture QC, and drug development process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
240338
2-Pyrrolidinone, ≥99%
C4H7NO
P74370
2-Pyrrolidinone, 99%
C4H7NO