EMAIL THIS PAGE TO A FRIEND
Stem cells and development
Lack of Rybp in Mouse Embryonic Stem Cells Impairs Cardiac Differentiation.
PMID 26110923
Abstract
Ring1 and Yy1 binding protein (Rybp) has been implicated in transcriptional regulation, apoptotic signaling and as a member of the polycomb repressive complex 1, it has an important function in regulating pluripotency and differentiation of embryonic stem cells (ESCs). Earlier, we had proved that Rybp plays an essential role in mouse embryonic and central nervous system development. This work identifies Rybp, as a critical regulator of heart development. Rybp is readily detectable in the developing mouse heart from day 8.5 of embryonic development. Prominent Rybp expression persists during all embryonic stages, and Rybp marks differentiated cell types of the heart. By utilizing rybp null ESCs in an in vitro cardiac differentiation assay, we found that rybp null ESCs do not form rhythmically beating cardiomyocytes (CMCs). Gene expression profiles revealed a downregulation of cardiac terminal and upregulation of germline-specific markers in the rybp null CMCs. Furthermore, transcriptome analysis uncovered a number of novel candidate target genes regulated by Rybp. Among these are several that are important in cardiac development and contractility such as Plagl1, Isl1, and Tnnt2. Importantly, forced expression of rybp in rybp-deficient ESCs by a lentiviral vector was able to rescue the mutant phenotype. Our data provide evidence for a previously unrecognized function of Rybp in heart development and point out the importance of germ cell lineage gene silencing during somatic differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3316
Anti-DEDAF antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
PRS2227
Anti-DEDAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501454
Anti-POU5F1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100956
Anti-POU5F1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101850
Anti-POU5F1 antibody produced in rabbit, affinity isolated antibody
SAB3500316
Anti-POU5F1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701958
Anti-POU5F1 antibody produced in rabbit
SAB2701960
Anti-POU5F1 antibody produced in rabbit
SAB2701972
Anti-POU5F1 antibody produced in rabbit
SAB1407211
Anti-RYBP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38973
Anti-RYBP antibody produced in rabbit, affinity isolated antibody
SAB2104016
Anti-RYBP antibody produced in rabbit, affinity isolated antibody
HPA053357
Anti-RYBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410422
Anti-RYBP antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103506
Anti-RYBP, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1404228
Monoclonal Anti-POU5F1, (C-terminal) antibody produced in mouse, clone 4F8, purified immunoglobulin, buffered aqueous solution
SAB1403139
Monoclonal Anti-RYBP antibody produced in mouse, clone 1F4, purified immunoglobulin, buffered aqueous solution