EMAIL THIS PAGE TO A FRIEND
Nature communications
Structure and gating of the nuclear pore complex.
PMID 26112706
Abstract
Nuclear pore complexes (NPCs) perforate the nuclear envelope and allow the exchange of macromolecules between the nucleus and the cytoplasm. To acquire a deeper understanding of this transport mechanism, we analyse the structure of the NPC scaffold and permeability barrier, by reconstructing the Xenopus laevis oocyte NPC from native nuclear envelopes up to 20 Å resolution by cryo-electron tomography in conjunction with subtomogram averaging. In addition to resolving individual protein domains of the NPC constituents, we propose a model for the architecture of the molecular gate at its central channel. Furthermore, we compare and contrast this native NPC structure to one that exhibits reduced transport activity and unveil the spatial properties of the NPC gate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
L1293
Anti-Lamin A (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4501766
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501767
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501764
Anti-Lamin A antibody produced in rabbit, affinity isolated antibody
SAB1406087
Anti-LMNA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006660
Anti-LMNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502100
Anti-LMNB1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502101
Anti-LMNB1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2101352
Anti-LMNB1 antibody produced in rabbit, affinity isolated antibody
SAB1410759
Anti-LMNB1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
M2643
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
MgSO4
203726
Magnesium sulfate, ≥99.99% trace metals basis
MgSO4
M3409
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
MgSO4
83266
Magnesium sulfate solution, BioUltra, for molecular biology
MgSO4
WH0000060M1
Monoclonal Anti-ACTB antibody produced in mouse, clone 3G4-F9, purified immunoglobulin, buffered aqueous solution
SAB1403520
Monoclonal Anti-ACTB antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
SAB4200236
Monoclonal Anti-Lamin A/C antibody produced in mouse, clone 4C11, purified from hybridoma cell culture
SAB1400153
Monoclonal Anti-LMNB1 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1404027
Monoclonal Anti-LMNB1 antibody produced in mouse, clone 4E4, purified immunoglobulin, buffered aqueous solution