PloS one
Mouse Tafazzin Is Required for Male Germ Cell Meiosis and Spermatogenesis.
PMID 26114544
Abstract
Barth syndrome is an X-linked mitochondrial disease, symptoms of which include neutropenia and cardiac myopathy. These symptoms are the most significant clinical consequences of a disease, which is increasingly recognised to have a variable presentation. Mutation in the Taz gene in Xq28 is thought to be responsible for the condition, by altering mitochondrial lipid content and mitochondrial function. Male chimeras carrying a targeted mutation of Taz on their X-chromosome were infertile. Testes from the Taz knockout chimeras were smaller than their control counterparts and this was associated with a disruption of the progression of spermatocytes through meiosis to spermiogenesis. Taz knockout ES cells also showed a defect when differentiated to germ cells in vitro. Mutant spermatocytes failed to progress past the pachytene stage of meiosis and had higher levels of DNA double strand damage and increased levels of endogenous retrotransposon activity. Altogether these data revealed a novel role for Taz in helping to maintain genome integrity in meiosis and facilitating germ cell differentiation. We have unravelled a novel function for the Taz protein, which should contribute to an understanding of how a disruption of the Taz gene results in the complex symptoms underlying Barth Syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA021803
Anti-ACTL7B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB2500296
Anti-DAZL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA019593
Anti-DAZL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019777
Anti-DAZL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2100531
Anti-DAZL antibody produced in rabbit, affinity isolated antibody
SAB1407479
Anti-HOOK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018537
Anti-HOOK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA018820
Anti-HOOK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB1410920
Anti-HOOK1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2107601
Anti-HPRT antibody produced in rabbit, affinity isolated antibody
SAB4200291
Anti-HPRT1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200292
Anti-HPRT1 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2701812
Anti-hprt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA006360
Anti-HPRT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV48471
Anti-HPRT1 antibody produced in rabbit, affinity isolated antibody
AV53820
Anti-PGK2 antibody produced in rabbit, affinity isolated antibody
SAB2700972
Anti-PGK2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
T5076
Monoclonal Anti-β-Tubulin Isotype III antibody produced in mouse, clone SDL.3D10, purified from hybridoma cell culture
SAB1400552
Monoclonal Anti-HOOK1 antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
WH0003251M1
Monoclonal Anti-HPRT1 antibody produced in mouse, clone 4C3-G8, purified immunoglobulin, buffered aqueous solution
SAB5300163
Monoclonal Anti-HPRT1 antibody produced in mouse, clone 1F8D11, ascites fluid