EMAIL THIS PAGE TO A FRIEND
PLoS pathogens
Fungal Morphology, Iron Homeostasis, and Lipid Metabolism Regulated by a GATA Transcription Factor in Blastomyces dermatitidis.
PMID 26114571
Abstract
In response to temperature, Blastomyces dermatitidis converts between yeast and mold forms. Knowledge of the mechanism(s) underlying this response to temperature remains limited. In B. dermatitidis, we identified a GATA transcription factor, SREB, important for the transition to mold. Null mutants (SREBΔ) fail to fully complete the conversion to mold and cannot properly regulate siderophore biosynthesis. To capture the transcriptional response regulated by SREB early in the phase transition (0-48 hours), gene expression microarrays were used to compare SREB∆ to an isogenic wild type isolate. Analysis of the time course microarray data demonstrated SREB functioned as a transcriptional regulator at 37°C and 22°C. Bioinformatic and biochemical analyses indicated SREB was involved in diverse biological processes including iron homeostasis, biosynthesis of triacylglycerol and ergosterol, and lipid droplet formation. Integration of microarray data, bioinformatics, and chromatin immunoprecipitation identified a subset of genes directly bound and regulated by SREB in vivo in yeast (37°C) and during the phase transition to mold (22°C). This included genes involved with siderophore biosynthesis and uptake, iron homeostasis, and genes unrelated to iron assimilation. Functional analysis suggested that lipid droplets were actively metabolized during the phase transition and lipid metabolism may contribute to filamentous growth at 22°C. Chromatin immunoprecipitation, RNA interference, and overexpression analyses suggested that SREB was in a negative regulatory circuit with the bZIP transcription factor encoded by HAPX. Both SREB and HAPX affected morphogenesis at 22°C; however, large changes in transcript abundance by gene deletion for SREB or strong overexpression for HAPX were required to alter the phase transition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379409
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone, 98%
C7H9NO2
266620
Boron, crystalline, 1 cm, 99.7% trace metals basis
B
333360
Boron, crystalline, −60 mesh, 99% trace metals basis
B
332445
Boron, amorphous, powder, (submicron)
B
15580
Boron, ≥95% (boron), amorphous powder
B
GF84487674
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF19976080
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF81914834
Boron, monofilament, 100m, diameter 0.1mm
B
GF54382888
Boron, monofilament, 10m, diameter 0.1mm
B
GF74687080
Boron, monofilament, 10m, diameter 0.2mm
B
GF40338132
Boron, monofilament, 5m, diameter 0.2mm
B
GF97861247
Boron, monofilament, 20m, diameter 0.1mm
B
GF04144586
Boron, monofilament, 20m, diameter 0.2mm
B
GF32655315
Boron, monofilament, 50m, diameter 0.1mm
B
GF70375833
Boron, monofilament, 50m, diameter 0.2mm
B
GF98835069
Boron, monofilament, 5m, diameter 0.1mm
B
GF39354048
Boron, monofilament, 200m, diameter 0.1mm
B
P6125
Pentadecanoic acid, ~99% (capillary GC)
C15H30O2
W433400
Pentadecanoic acid, 99%
C15H30O2
S4751
Stearic acid, Grade I, ≥98.5% (capillary GC)
C18H36O2
W303518
Stearic acid, ≥95%, FCC, FG
C18H36O2
PZ0005
Voriconazole, ≥98% (HPLC)
C16H14F3N5O