EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
The home-made in situ passive flux sampler for the measurement of monoterpene emission flux: preliminary studies.
PMID 26116240
Abstract
The paper presents the construction and metrological characteristics of the home-made in situ passive flux sampler, an analytical tool representing small-scale emission chambers working in situ and passively sampling analytes from the gaseous phase. The sorption element was a cylindrical container made of stainless steel net, packed with a carbon sorbent bed-graphitized charcoal, Carbograph 4 (35/50xa0mesh). The recommended working/exposure time of the constructed passive device was determined by carrying out model tests in the laboratory. In addition, a preliminary study was conducted to determine the rate of the emission flux of selected monoterpenes released from the surface of wood-based indoor materials (laminated chipboard) used in residential areas.