EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Grape marc as a source of carbohydrates for bioethanol: Chemical composition, pre-treatment and saccharification.
PMID 26117238
Abstract
Global grape production could generate up to 13 Mt/yr of wasted biomass. The compositions of Cabernet Sauvignon (red marc) and Sauvignon Blanc (white marc) were analyzed with a view to using marc as raw material for biofuel production. On a dry weight basis, 31-54% w/w of the grape marc consisted of carbohydrate, of which 47-80% was soluble in aqueous media. Ethanol insoluble residues consisted mainly of polyphenols, pectic polysaccharides, heteroxylans and cellulose. Acid and thermal pre-treatments were investigated for their effects on subsequent cellulose saccharification. A 0.5M sulfuric acid pre-treatment yielded a 10% increase in the amount of liberated glucose after enzymatic saccharification. The theoretical amount of bioethanol that could be produced by fermentation of grape marc was up to 400 L/t. However, bioethanol from only soluble carbohydrates could yield 270 L/t, leaving a polyphenol enriched fraction that may be used in animal feed or as fertilizer.