EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Butyltins, polyaromatic hydrocarbons, organochlorine pesticides, and polychlorinated biphenyls in sediments and bivalve mollusks in a mid-latitude environment from the Patagonian coastal zone.
PMID 26118658
Abstract
Butyltins (BTs), polyaromatic hydrocarbons (PAHs), organochlorine pesticides (OCPs), and polychlorinated biphenyls (PCBs) were assessed in a mid-latitude environment of the Patagonian coast, distant from significant pollutant sources. Bioaccumulation processes through bottom sediment resuspension were suggested by BTs level (expressed as ng of tin [Sn] g(-1) dry wt) found in surface sediment (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48508
Hexachlorobenzene, analytical standard
C6Cl6