EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Development and validation of an UPLC-MS/MS method for the quantification of tamoxifen and its main metabolites in human scalp hair.
PMID 26119504
Abstract
The aim of this study was to validate an earlier developed high-performance highly sensitive ultra performance liquid chromatography/tandem mass spectrometry (UPLC-MS/MS) method for quantification of tamoxifen and its three main metabolites (N-desmethyl-tamoxifen, 4-hydroxy-tamoxifen and 4-hydroxy-N-desmethyl-tamoxifen) in scalp hair. This non-invasive method might, by segmental analysis of hair, be useful in the determination of the concentration of drugs and its metabolites over time, which can be used to study a wide variety of clinical relevant questions. Hair samples (150-300 hair strands, cut as close to the scalp as possible from the posterior vertex region of the head) were collected from female patients taking tamoxifen 20mg daily (n=19). The analytes were extracted using a liquid-liquid extraction procedure with carbonate buffer at pH 8.8 and a mixture of n-hexane/isopropranol method, followed by UPLC-MS/MS chromatography, based on an earlier validated method. The calibration curves were linear in the range of 1.00-200 pmol for tamoxifen and N-desmethyl-tamoxifen, with lower limit of quantitation of 1.00 pmol and 0.100-20.0 pmol with lower limit of quantitation of 0.100 pmol for endoxifen and 4-hydroxy-tamoxifen. Assay performance was fair with a within-run and between-run variability less than 9.24 at the three quality control samples and less than 15.7 for the lower limit of quantitation. Importantly, a steep linear decline was observed from distal to proximal hair segments. Probably, this is due to UV exposure as we showed degradation of tamoxifen and its metabolites after exposure to UV-light. Furthermore, higher concentrations of tamoxifen were found in black hair samples compared to blond and brown hair samples. We conclude that measurement of the concentration of tamoxifen and its main metabolites in hair is possible, with the selective, sensitive, accurate and precise UPLC-MS/MS method. However, for tamoxifen, it seems not possible to determine exposure over time with segmental analysis of hair, probably largely due to the effect of UV irradiation. Further research should therefore focus on quantification of other anticancer drugs, in segmented scalp hair, that are less sensitive to UV irradiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS013577
ammonium carbonate, AldrichCPR
C2H13N3O6
68392
Ammonium carbonate, tested according to NF
CH8N2O3