EMAIL THIS PAGE TO A FRIEND
Journal of applied microbiology
Lactobacillus kunkeei YB38 from honeybee products enhances IgA production in healthy adults.
PMID 26121394
Abstract
To identify lactic acid bacterial isolates, which promote immunoglobulin A (IgA) production in honeybee products and honeybees (Apis mellifera). Pyrosequencing analysis of the microbiota of honeybee products and honeybees revealed the predominance of Lactobacillus kunkeei in honey, bee pollen, bee bread and royal jelly. Lactobacillus kunkeei was isolated from bee pollen, bee bread and honey stomach, and its effect on IgA production was evaluated in vitro. Heat-killed YB38 and YB83 isolates from bee pollen promoted IgA production in mouse Peyer's Patch cells and had little mitogenic activity or effect on IL-2 production in mouse spleen cells in comparison with Listeria monocytogenes, which does exhibit mitogen activity. A pilot study in 11 healthy adults showed that 4-week intake of 1000 mg day(-1) heat-killed YB38 increased secretory IgA (SIgA) concentrations and secretion in saliva with no adverse effects. Heat-killed Lact. kunkeei YB38 from bee pollen increases IgA production and may safely improve immune responsiveness. This is the first report of microbiota analysis of royal jelly and the immune efficacy of Lact. kunkeei from honeybee products in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9379 Lectin from Phytolacca americana (pokeweed), lyophilized powder