EMAIL THIS PAGE TO A FRIEND
Surgical laparoscopy, endoscopy & percutaneous techniques
Results of Immediate Discharge From Postanesthesia Care Unit to Home After Laparoscopic Appendectomy for Acute Nonperforated Appendicitis.
PMID 26121544
Abstract
This study examines the efficacy and safety of immediate discharge from the postanesthesia care unit after laparoscopic appendectomy for patients with acute nonperforated appendicitis. A retrospective study of 114 laparoscopic appendectomy cases was performed. Seventy-four patients met inclusion criteria of having uncomplicated acute appendicitis. Patient demographics, operative times, length of postoperative stay, and complication rates were analyzed. Only 3 of the 74 patients failed to be discharged immediately. Mean postoperative discharge time was 130.5 minutes with 86% of patients leaving within 3 hours. Delay in voiding was the most common cause of stays longer than 4 hours. Six of the 74 patients (8%) returned to the emergency room without need for readmission. There were no complications associated with the early discharge. Immediate discharge from the postanesthesia care unit for patients with acute nonperforated appendicitis appears safe and effective in the community hospital setting.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
399396
Benzoin, purified by sublimation, ≥99.5%
C14H12O2
B8681
Benzoin, 98%
C14H12O2