EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
MicroRNA-132 enhances transition from inflammation to proliferation during wound healing.
PMID 26121747
Abstract
Wound healing is a complex process that is characterized by an initial inflammatory phase followed by a proliferative phase. This transition is a critical regulatory point; however, the factors that mediate this process are not fully understood. Here, we evaluated microRNAs (miRs) in skin wound healing and characterized the dynamic change of the miRNome in human skin wounds. miR-132 was highly upregulated during the inflammatory phase of wound repair, predominantly expressed in epidermal keratinocytes, and peaked in the subsequent proliferative phase. TGF-β1 and TGF-β2 induced miR-132 expression in keratinocytes, and transcriptome analysis of these cells revealed that miR-132 regulates a large number of immune response- and cell cycle-related genes. In keratinocytes, miR-132 decreased the production of chemokines and the capability to attract leukocytes by suppressing the NF-κB pathway. Conversely, miR-132 increased activity of the STAT3 and ERK pathways, thereby promoting keratinocyte growth. Silencing of the miR-132 target heparin-binding EGF-like growth factor (HB-EGF) phenocopied miR-132 overexpression in keratinocytes. Using mouse and human ex vivo wound models, we found that miR-132 blockade delayed healing, which was accompanied by severe inflammation and deficient keratinocyte proliferation. Together, our results indicate that miR-132 is a critical regulator of skin wound healing that facilitates the transition from the inflammatory to the proliferative phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-182
Anti-MAP Kinase 1/2 (Erk1/2) Antibody, CT, 100 µg, Anti-MAP Kinase 1/2 (Erk1/2) Antibody, CT detects level of MAP Kinase 1/2 (Erk1/2) & has been published & validated for use in IC, IP & WB.
SAB2104809
Anti-EGF antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700260 Anti-goat IgG (Fc specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700261 Anti-goat IgG (Fc specific)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700258 Anti-goat IgG (Fc specific)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700259 Anti-goat IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700256 Anti-goat IgG (Fc specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700262 Anti-goat IgG (Fc specific)-Texas Red® antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4600053
Anti-Goat IgG (H+L), CF 488A antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600072
Anti-Goat IgG (H+L), CF 555 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600088
Anti-Goat IgG (H+L), CF 568 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600118
Anti-Goat IgG (H+L), CF 594 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600148
Anti-Goat IgG (H+L), CF 633 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600171
Anti-Goat IgG (H+L), CF 640R antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600188
Anti-Goat IgG (H+L), CF 647 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600203
Anti-Goat IgG (H+L), CF 680 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB3700304 Anti-goat IgG (H+L), F(ab′)2 fragment-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700305 Anti-goat IgG (H+L), F(ab′)2 fragment-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700302 Anti-goat IgG (H+L), F(ab′)2 fragment-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700303 Anti-goat IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700299 Anti-goat IgG (H+L), F(ab′)2 fragment-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700306 Anti-goat IgG (H+L), F(ab′)2 fragment-Texas Red® antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700325 Anti-Goat IgG (H+L), F(ab) fragment-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700323 Anti-Goat IgG (H+L), F(ab) fragment-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700324 Anti-Goat IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700321 Anti-Goat IgG (H+L), F(ab) fragment-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700326 Anti-Goat IgG (H+L), F(ab) fragment-Texas Red® antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700270 Anti-goat IgG (H+L)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
A9294 Anti-Goat IgG (whole molecule)−Agarose antibody produced in rabbit, IgG fraction of antiserum, PBS suspension
A4187 Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution