EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
IL-34 mediates acute kidney injury and worsens subsequent chronic kidney disease.
PMID 26121749
Abstract
Macrophages (Mø) are integral in ischemia/reperfusion injury-incited (I/R-incited) acute kidney injury (AKI) that leads to fibrosis and chronic kidney disease (CKD). IL-34 and CSF-1 share a receptor (c-FMS), and both cytokines mediate Mø survival and proliferation but also have distinct features. CSF-1 is central to kidney repair and destruction. We tested the hypothesis that IL-34-dependent, Mø-mediated mechanisms promote persistent ischemia-incited AKI that worsens subsequent CKD. In renal I/R, the time-related magnitude of Mø-mediated AKI and subsequent CKD were markedly reduced in IL-34-deficient mice compared with controls. IL-34, c-FMS, and a second IL-34 receptor, protein-tyrosine phosphatase ζ (PTP-ζ) were upregulated in the kidney after I/R. IL-34 was generated by tubular epithelial cells (TECs) and promoted Mø-mediated TEC destruction during AKI that worsened subsequent CKD via 2 distinct mechanisms: enhanced intrarenal Mø proliferation and elevated BM myeloid cell proliferation, which increases circulating monocytes that are drawn into the kidney by chemokines. CSF-1 expression in TECs did not compensate for IL-34 deficiency. In patients, kidney transplants subject to I/R expressed IL-34, c-FMS, and PTP-ζ in TECs during AKI that increased with advancing injury. Moreover, IL-34 expression increased, along with more enduring ischemia in donor kidneys. In conclusion, IL-34-dependent, Mø-mediated, CSF-1 nonredundant mechanisms promote persistent ischemia-incited AKI that worsens subsequent CKD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV45673
Anti-ARG1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501375
Anti-ARG1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405480
Anti-ARG1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003595
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024006
Anti-ARG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100144
Anti-ARG1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4200510
Anti-ARG1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1406413
Anti-CCL2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019163
Anti-CCL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103318
Anti-CCL2 antibody produced in rabbit, affinity isolated antibody
SAB1411231
Anti-CCL2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA022132
Anti-CD38 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1301488
ANTI-CLEC7A (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA043244
Anti-CLEC7A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500794
Anti-CLEC7A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV44329
Anti-CSF1 antibody produced in rabbit, affinity isolated antibody
SAB1410248
Anti-CSF1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3701329 Anti-Human IgG (H+L) antibody produced in mouse, affinity isolated antibody, buffered aqueous solution
PRS4779
Anti-IL-34 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4781
Anti-IL-34 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
F6258 Anti-Rat IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
RAB0699 Human IL34 / Interleukin-34 ELISA Kit
SAB4200508
Monoclonal Anti-ARG1 antibody produced in mouse, ~1.0 mg/mL, clone ARG1-3, purified immunoglobulin
SAB4100251
Monoclonal Anti-ARG1 antibody produced in mouse, clone ARG1-A219, culture supernatant
SAB4200509
Monoclonal Anti-ARG1 antibody produced in mouse, ~1.0 mg/mL, clone ARG1-6, purified immunoglobulin
AMAB90545
Monoclonal Anti-ARG1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0186, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402908
Monoclonal Anti-CCL2 antibody produced in mouse, clone 1F10, purified immunoglobulin, buffered aqueous solution
SAB1409632
Monoclonal Anti-CCL2 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
WH0000952M2
Monoclonal Anti-CD38 antibody produced in mouse, clone 4G3, purified immunoglobulin, buffered aqueous solution
SAB4700170 Monoclonal Anti-CD38 antibody produced in mouse, clone HIT2, purified immunoglobulin, buffered aqueous solution