EMAIL THIS PAGE TO A FRIEND
BMC research notes
Evidence that histone H1 is dispensable for proper meiotic recombination in budding yeast.
PMID 26122007
Abstract
Histone H1, referred to as the linker histone, associates with the nucleosome core particle. While there is indication that the budding yeast version of histone H1 (Hho1) contributes to regulation of chromatin structure and certain chromatin-related processes, such as DNA double-strand break repair, cells lacking Hho1 are healthy and display subtle phenotypes. A recent report has revealed that Hho1 is required for optimal sporulation. The studies described here were conducted to determine whether Hho1 influences meiotic recombination, an event that occurs during sporulation, involves generation and repair of DNA double-strand breaks, and is critical for spore viability. Through tetrad analysis, cells with or without Hho1 were compared for meiotic reciprocal recombination events within several chromosome XV intervals. Parameters investigated included crossover frequency (genetic map distance) and crossover interference. No significant differences were detected between the two cell types. In agreement with earlier studies, spore viability was not affected by Hho1 absence. These data suggest that complete absence of Hho1 from chromatin does not affect reciprocal recombination between homologous chromosomes during meiosis. Therefore, the basal level of Hho1 that remains after its reported depletion early in meiosis is unlikely to be important for regulating recombination. Furthermore, the subsequent accumulation of Hho1 as the haploid products mature does not appear to be crucial for spore viability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3553
Acrylamide, for electrophoresis, ≥99% (HPLC), powder
C3H5NO
A8887
Acrylamide, suitable for electrophoresis, ≥99%
C3H5NO
A9099
Acrylamide, for molecular biology, ≥99% (HPLC)
C3H5NO
01698
Acrylamide, for electrophoresis, ≥99.0% (GC)
C3H5NO
A4058
Acrylamide solution, 40%, for electrophoresis, sterile-filtered
C3H5NO
W302902
D-Sorbitol, ≥98%, FCC, FG
C6H14O6
240850
D-Sorbitol, 99%
C6H14O6
S1876
D-Sorbitol, ≥98%
C6H14O6
85529
D-Sorbitol, BioUltra, ≥99.5% (HPLC)
C6H14O6
S6021
D-Sorbitol, for molecular biology, ≥98%
C6H14O6
S7547
D-Sorbitol, BioXtra, ≥98%
C6H14O6
S3889
D-Sorbitol, BioReagent, cell culture tested, plant cell culture tested
C6H14O6
S0900
D-Sorbitol, for electrophoresis
C6H14O6
S8143
D-Sorbitol, plant cell culture tested
C6H14O6
E8751
Ethidium bromide, ~95% (HPLC)
C21H20BrN3
E7637
Ethidium bromide, BioReagent, for molecular biology, powder
C21H20BrN3
46065
Ethidium bromide, for fluorescence, ≥95.0% (HPLC)
C21H20BrN3
E1510
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
C21H20BrN3
E1385
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
C21H20BrN3
46067
Ethidium bromide solution, for fluorescence, ~1% in H2O
C21H20BrN3
517992
Lithium acetate, 99.95% trace metals basis
C2H3LiO2
146072
N,N′-Methylenebis(acrylamide), 99%
C7H10N2O2
M7279
N,N′-Methylenebisacrylamide, powder, for molecular biology, for electrophoresis, ≥99.5%
C7H10N2O2
M7256
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
C7H10N2O2
M1533
N,N′-Methylenebisacrylamide solution, for electrophoresis, 2% in H2O
C7H10N2O2
46047
Pyridaben, PESTANAL®, analytical standard
C19H25ClN2OS
190780
Sodium trichloroacetate, 97%
C2Cl3NaO2
309532
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
C6H10O6