EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology
Cold acclimation increases levels of some heat shock protein and sirtuin isoforms in threespine stickleback.
PMID 26123780
Abstract
Molecular chaperones [heat shock proteins (HSPs)] increase in response to rapid changes in temperatures, but long-term acclimation to cold temperature may also warrant elevations in HSPs. In fishes, cold acclimation increases mitochondrial density and oxidative stress in some tissues, which may increase demand for HSPs. We hypothesized that levels of HSPs, as well as sirtuins (SIRTs), NAD-dependent deacetylases that mediate changes in metabolism and responses to oxidative stress (including increases in HSPs), would increase during cold acclimation of threespine stickleback (Gasterosteus aculeatus). Transcript levels of hsp70, hsc70, hsp60 and hsp90-α, sirts1-4, as well as protein levels of HSP60, HSP90 and HSC70 were quantified in liver and pectoral adductor muscle of stickleback during cold acclimation from 20 °C to 8 °C. In liver, cold acclimation stimulated a transient increase in mRNA levels of hsp60 and hsc70. Transcript levels of sirt1 and sirt2 also increased in response to cold acclimation and remained elevated. In pectoral muscle, mRNA levels of hsp60, hsp90-α, hsc70 and sirt1 all transiently increased in response to cold acclimation, while levels of sirts2-4 remained constant or declined. Similar to transcript levels, protein levels of HSC70 increased in both liver and pectoral muscle. Levels of HSP90 also increased in liver after 4 weeks at 8 °C. HSP60 remained unchanged in both tissues, as did HSP90 in pectoral muscle. Our results indicate that while both HSPs and SIRTs increase in response to cold acclimation in stickleback, the response is tissue and isoform specific, likely reflecting differences in metabolism and oxidative stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z278343 ACE Micro/Mini-Lab® kits conical funnel, top diam. 25 mm, tube diam. × L 5 mm × 20 mm
Z174858
Adapter for ACE Micro/Mini-Lab® kit, Distilling, Joint: ST/NS 14/10
Z175013
Adapter for ACE Micro/Mini-Lab® kit, Mini adapter
Z174874
Adapter for ACE Micro/Mini-Lab® kit, Vacuum-takeoff with hose barb, Joint: ST/NS 14/10
Z288497
Aldrich® Imperial mini-spill kits, capacity 1.8 gal (6.8 L)
Z288500
Aldrich® Imperial mini-spill kits, capacity 3.6 gal (13.6 L)
SAB4501464
Anti-HSP60, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2701704
Anti-hspa8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701706
Anti-hspa8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701813
Anti-hspa8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701964
Anti-HSPA8 antibody produced in rabbit
SAB2702050
Anti-HSPA8 antibody produced in rabbit
SAB2702054
Anti-HSPA8 antibody produced in rabbit
SAB2104919
Anti-HSPA8, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1405973
Anti-HSPD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001523
Anti-HSPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107604
Anti-HSPD1 antibody produced in rabbit, affinity isolated antibody
SAB1410668
Anti-HSPD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
H3524 Monoclonal Anti-Heat Shock Protein 60 antibody produced in mouse, clone LK2, ascites fluid
H4149 Monoclonal Anti-Heat Shock Protein 60 antibody produced in mouse, clone LK1, ascites fluid
SAB1412719
Monoclonal Anti-HSPA8 antibody produced in mouse, clone 1G2, purified immunoglobulin, 1 mg/mL
SAB4100596
Monoclonal Anti-HSPD1 antibody produced in mouse, clone HSPD1-A580, culture supernatant
SAB4100597
Monoclonal Anti-HSPD1 antibody produced in mouse, clone HSPD1-B580, culture supernatant
PLA0269
Rabbit anti-HSP60 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.