EMAIL THIS PAGE TO A FRIEND
Molecular & cellular proteomics : MCP
Quantitative Proteomics Reveals the Roles of Peroxisome-associated Proteins in Antiviral Innate Immune Responses.
PMID 26124285
Abstract
Compared with whole-cell proteomic analysis, subcellular proteomic analysis is advantageous not only for the increased coverage of low abundance proteins but also for generating organelle-specific data containing information regarding dynamic protein movement. In the present study, peroxisome-enriched fractions from Sendai virus (SeV)-infected or uninfected HepG2 cells were obtained and subjected to quantitative proteomics analysis. We identified 311 proteins that were significantly changed by SeV infection. Among these altered proteins, 25 are immune response-related proteins. Further bioinformatic analysis indicated that SeV infection inhibits cell cycle-related proteins and membrane attack complex-related proteins, all of which are beneficial for the survival and replication of SeV within host cells. Using Luciferase reporter assays on several innate immune-related reporters, we performed functional analysis on 11 candidate proteins. We identified LGALS3BP and CALU as potential negative regulators of the virus-induced activation of the type I interferons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
40291
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)
H5NO
40321
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade SLSI PURANAL (Honeywell 17301)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
SAB2500013
Anti-ABCD3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA032027
Anti-ABCD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS2301
Anti-AIF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500441
Anti-AIF antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406783
Anti-AIFM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100079
Anti-AIFM1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA030611
Anti-AIFM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405551
Anti-CALU antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006018
Anti-CALU antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1307018
ANTI-CALU(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406082
Anti-LGALS3BP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000554
Anti-LGALS3BP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV54779
Anti-LGALS3BP antibody produced in rabbit, affinity isolated antibody
SAB2701961
Anti-LGALS3BP antibody produced in rabbit
AV40199
Anti-PDCD8 (AB1) antibody produced in rabbit, affinity isolated antibody
AV00028
Anti-PDCD8 (AB2) antibody produced in rabbit, affinity isolated antibody
P0497
Anti-PMP70 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200181
Anti-PMP70 antibody, Mouse monoclonal, clone 70-18, purified from hybridoma cell culture
SAB1402888
Monoclonal Anti-ABCD3 antibody produced in mouse, clone 2H6, purified immunoglobulin, buffered aqueous solution
SAB5300151
Monoclonal Anti-AIFM1 antibody produced in mouse, clone 4E7, ascites fluid
H9658
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
RDD003
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C7H15NO4S
M1254
MOPS, ≥99.5% (titration)
C7H15NO4S