EMAIL THIS PAGE TO A FRIEND
The Biological bulletin
Dual Cellular Supporters: Multi-Layer Glial Wrapping and the Penetrative Matrix Specialized in Deep-Sea Hydrothermal Vent Endemic Scale-Worms.
PMID 26124448
Abstract
Hydrothermal vent organisms undergo extreme environments that may require unique innovations. The present study reports a distinct case of cellular supportive systems in the nervous systems of a scale-worm, Branchinotogluma japonica, endemic to deep-sea hydrothermal vents. We found two organizations in the tissues of these animals. First, multi-layers of glia ensheath the ventral cell bodies of the brain and ventral nerve cord, in a manner similar to that of myelin or lamellar ensheathments. Second, matrices of numerous penetrative fibers, or tonofilaments, composed of bundles of ca. 20-nm fibers, are directly connected with the basal parts of epidermal cuticles and run into the diffuse intercellular spaces of the brain neuropils and peripheral nerves. Both types of tissue might be mechanical supportive structures for the neuronal cell bodies. In addition, as a glial function, the multi-layer membranes and the epithelial support cells may be required for physicochemical homeostatic regulation to filter toxic heavy metals and for inhibiting breakdown of glial membrane integrity under strong oxidative stress imposed by hypoxia in the hydrothermal vent environment. Similar functions are known in the well-studied cases of the blood-brain barrier in mammalian brains, including in human stroke.