EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Real-Time Monitoring of Cellular Bioenergetics with a Multianalyte Screen-Printed Electrode.
PMID 26125545
Abstract
Real-time monitoring of changes to cellular bioenergetics can provide new insights into mechanisms of action for disease and toxicity. This work describes the development of a multianalyte screen-printed electrode for the detection of analytes central to cellular bioenergetics: glucose, lactate, oxygen, and pH. Platinum screen-printed electrodes were designed in-house and printed by Pine Research Instrumentation. Electrochemical plating techniques were used to form quasi-reference and pH electrodes. A Dimatix materials inkjet printer was used to deposit enzyme and polymer films to form sensors for glucose, lactate, and oxygen. These sensors were evaluated in bulk solution and microfluidic environments, and they were found to behave reproducibly and possess a lifetime of up to 6 weeks. Linear ranges and limits of detection for enzyme-based sensors were found to have an inverse relationship with enzyme loading, and iridium oxide pH sensors were found to have super-Nernstian responses. Preliminary measurements where the sensor was enclosed within a microfluidic channel with RAW 264.7 macrophages were performed to demonstrate the sensors' capabilities for performing real-time microphysiometry measurements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1472
Potassium carbonate, meets USP testing specifications
CK2O3
367877
Potassium carbonate, 99.995% trace metals basis
CK2O3
590681
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
CK2O3
60108
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
CK2O3
P9333
Potassium chloride, BioXtra, ≥99.0%
ClK
P9541
Potassium chloride, for molecular biology, ≥99.0%
ClK
P5405
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
ClK
P1597
Potassium chloride, meets EP, USP testing specifications
ClK
449989
Potassium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClK
451029
Potassium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClK
409316
Potassium chloride, 99.999% trace metals basis
ClK
204099
Potassium chloride, ≥99.99% trace metals basis
ClK
60128
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClK
P9327
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
ClK
60142
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
ClK
44675
Potassium chloride solution, 1 M
ClK
L7022
Sodium L-lactate, ~98%
C3H5NaO3
71718
Sodium L-lactate, ≥99.0% (NT)
C3H5NaO3
339741
Sulfuric acid, 99.999%
H2O4S
28-5930
Sulfuric acid solution, 1 M
H2O4S